Skip to Content
Merck
All Photos(2)

Key Documents

MABF1110

Sigma-Aldrich

Anti-UBR5 Antibody, clone 16G10.1

clone 16G10.1, from mouse

Synonym(s):

E3 ubiquitin-protein ligase UBR5, EC: 2.3.2.26, E3 ubiquitin-protein ligase, HECT domain-containing 1, HECT-type E3 ubiquitin transferase UBR5, Hyperplastic discs protein homolog, hHYD, Progestin-induced protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€341.00

€341.00


Estimated to ship on28 May 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1698

Request a Bulk Order

Select a Size

Change View
100 μG
€341.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€341.00


Estimated to ship on28 May 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1698

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

16G10.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... UBR5(51366)

Related Categories

General description

E3 ubiquitin-protein ligase UBR5 (UniProt; O95071; also known as EC: 2.3.2.26, E3 ubiquitin-protein ligase, HECT domain-containing 1, HECT-type E3 ubiquitin transferase UBR5, Hyperplastic discs protein homolog, hHYD, Progestin-induced protein) is encoded by the UBR5 (also known as EDD, EDD1, HYD, KIAA0896) gene (Gene ID: 51366) in human. UBR5 is a widely expressed nuclear E3 ligase that is most abundant in testis and expressed at high levels in brain, pituitary and kidney. It is a component of the N-end rule pathway that recognizes and binds to proteins bearing specific N-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation. UBR5 is involved in maturation and/or transcriptional regulation of mRNA by activating CDK9 by polyubiquitination and may play a role in control of cell cycle progression. It is shown to regulate DNA topoisomerase II binding protein (TopBP1) in the DNA damage response and may have a tumor suppressive function. It acts as a suppressor of RNF168, an E3 ubiquitin-protein ligase that promotes accumulation of Lys-63-linked histone H2A and H2AX at DNA damage sites, thereby acting as a guard against excessive spreading of ubiquitinated chromatin at damaged chromosomes. URB5 contains a zinc-finger domain (aa 1177-1245), a poly(A)-binding protein C-terminal (PABC) domain(aa 2377-2454), and a (Homologous to the E6-AP Carboxyl Terminus (HECT) domain (aa 2462-2799). Two isoforms of UBR5 have been described that are produced by alternative splicing.

Specificity

Clone 16G10.1 detects E3 ubiquitin-protein ligage UBR5 in human cells. It targets an epitope within 181 amino acids from the C-terminal half.

Immunogen

GST/His-tagged recombinant fragment corresponding to 181 amino acids from the C-terminal half of human E3 ubiquitin-protein ligase UBR5.

Application

Anti-UBR5, clone 16G10.1, Cat. No. MABF1110, is a mouse monoclonal antibody that detects E3 ubiquitin-protein ligase UBR5 and is tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected UBR5 in human cerebral cortex, human tonsil, and human prostate cancer tissues.
Research Category
Inflammation & Immunology

Quality

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected UBR5 in 10 µg of Raji cell lysate.

Target description

~305 kDa oserved; 309.35 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neel Dhingani et al.
Scientific reports, 10(1), 18648-18648 (2020-10-31)
Very early onset inflammatory bowel disease (VEOIBD) denotes children with onset of IBD before six years of age. A number of monogenic disorders are associated with VEOIBD including tetratricopeptide repeat domain 7A (TTC7A) deficiency. TTC7A-deficiency is characterized by apoptotic colitis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service