Skip to Content
Merck
All Photos(2)

Key Documents

MABE455

Sigma-Aldrich

Anti-CHD-4 Antibody, clone 3F2/4

clone 3F2/4, from mouse

Synonym(s):

Chromodomain-helicase-DNA-binding protein 4, CHD-4, ATP-dependent helicase CHD4, Mi-2 autoantigen 218 kDa protein, Mi2-beta

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€484.00

€484.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€484.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€484.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F2/4, monoclonal

species reactivity

mouse, human

technique(s)

ChIP: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CHD4(1108)

General description

CHD-4, often referred to as Mi2-beta, is a central component of the nucleosome remodeling and histone deacetylase (NuRD) repressor complex. CHD-4 has been shown to interact with TRIM27, HDAC1, Histone deacetylase 2, MTA2, SATB1 and Ataxia telangiectasia and Rad3 related. This protein also interacts with KLF1. Sumoylation of KLF1 leads to transcriptional repression of CHD-4.

Immunogen

Linear peptide corresponding to human CHD-4.

Application

Detect Chd4 (Chromodomain-helicase-DNA-binding protein 4) using this mouse monoclonal antibody, Anti-CHD-4 Antibody, clone 3F2/4 validated for use in western blotting, IHC & Immunofluorescence.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected CHD-4 in human breast carcinoma and skin tissues.

Chromatin Immunoprecipitation Analysis: A representative lot from an independent laboratory detected CHD-4 in mouse embryonic endothelial cells.

Immunofluorescence Analysis: A representative lot from an independent laboratory detected CHD-4 in mouse embryonic endothelial cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected CHD-4 in 10 µg of HeLa cell lysate.

Target description

~220 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Shi et al.
Development (Cambridge, England), 151(8) (2024-04-15)
Regulation of chromatin states is essential for proper temporal and spatial gene expression. Chromatin states are modulated by remodeling complexes composed of components that have enzymatic activities. CHD4 is the catalytic core of the nucleosome remodeling and deacetylase (NuRD) complex

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service