Skip to Content
Merck
All Photos(3)

Key Documents

MABE443

Sigma-Aldrich

Anti-BPTF Antibody, clone 15B1.1

clone 15B1.1, from mouse

Synonym(s):

Protein Bptf, BPTF

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€395.00

€395.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€395.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€395.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

15B1.1, monoclonal

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BPTF(2186)
mouse ... Bptf(207165)
rat ... Bptf(303617)

General description

Nucleosome-remodeling factor (NURF) subunit BPTF (UniProt E9Q6A7 & A2A654; also known as Bromodomain and PHD finger-containing transcription factor, Fetal Alzheimer antigen) is encoded by the Bptf (also known as 9430093H17Rik, Falz) gene (Gene ID 207165) in murine species. ATP-dependent chromatin remodeling complexes are classified into four major subfamilies (SWI/SNF, ISWI, CHD, and INO80). NURF is the founding member of ISWI remodeling complexes. Mammalian NURF is composed of BPTF, the ATPase SNF2L, and the Trp-Asp (WD) repeat protein pRBAP46/48, with BPTF being the largest subunit among the three. BPTF PHD finger-mediated binding of H3K4me3 stabilizes the NURF complex on chromatin, resulting in NURF-catalyzed nucleosome sliding. The NURF remodeling complex is essential for mammalian development and executes epigenetic regulation in multiple signaling pathways, including heat shock, TGF-beta/SMAD, JAK/STAT, WNT/beta-catenin, RB, and nuclear hormone receptors.

Immunogen

GST-tagged recombinant protein corresponding to mouse BPTF.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected BPTF in human testis, rat colon, and mouse colon tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-BPTF Antibody, clone 15B1.1 is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of BPTF .

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected BPTF in 10 µg of HeLa cell lysate.

Target description

~322 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service