Skip to Content
Merck
All Photos(3)

Key Documents

MABE215

Sigma-Aldrich

Anti-Liver receptor homolog-1 Antibody, clone 1LRH-2E1

ascites fluid, clone 1LRH-2E1, from mouse

Synonym(s):

Nuclear receptor subfamily 5 group A member 2, Liver receptor homolog 1, LRH-1, NR5A2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€395.00

€395.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€395.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€395.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1LRH-2E1, monoclonal

species reactivity

mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Nr5A2(26424)

General description

Liver receptor homolog 1 (LRH-1, CPF, FTF, PHR-1, FTF, or hB1F) is an orphan nuclear receptor that is highly expressed in the intestines, pancreas, ovary, and liver. LRH-1 is a constitutively active transcription factor with a complex role in many cellular processes which may include lipid metabolism, cell proliferation, development, reproduction, and immunity. LRH-1 may interact with an array of transcriptional coactivators that modulate its activity, among which are the MBF1, SRC, and PGC-1α proteins. In addition, ERK phosphorylation on Ser-238 and Ser-243 also enhances LRH-1 activity. However, LRH-1 is also negatively regulated by several factors. For instance, corepressors such as SHP, DAX-1, and PROX-1, preclude its binding to DNA; and sumoylation promotes its inactivation and sequestration into nuclear bodies. LRH-1 targets many genes including the CYP7A1 and CYP8B1 genes involved in bile synthesis; and Oct4 required for pluripotency in embryonic stem cells. LRH-1 has been implicated in inflammatory bowel disease, and tumor development in the intestines and pancreas.

Immunogen

Linear peptide corresponding to the mouse Liver receptor homolog-1.

Application

Use Anti-LRH-1 Antibody, clone 1LRH-2E1 (Mouse Monoclonal Antibody) validated in WB, ICC to detect LRH-1 also known as Liver receptor homolog 1.
Western Blot Analysis: A 1:1,000 dilution from representative lot detected Liver receptor homolog-1 in HepG2 and HeLa cell lysates (M. Oulad, IGBMC, France.).

Immunocytochemistry Analysis: A representative lot detected Liver receptor homolog-1 in HepG2 and HeLa cells (M. Oulad, IGBMC, France.).

Quality

Evaluated by Western Blot in mouse pancreas tissue lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Liver receptor homolog-1 in 10 µg of mouse pancrease tissue lysate.

Target description

~64 kDa, ~56 kDa, and ~42 kDa observed. This antibody detects three isoforms of Liver receptor homolog-1 as described by Uniprot No. O00482.

Physical form

Mouse monoclonal IgG2aλ ascites with 0.05% sodium azide.

Analysis Note

Control
Mouse pancreas tissue lysate

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service