Skip to Content
Merck
All Photos(2)

Key Documents

MABE1982

Sigma-Aldrich

Anti-REST Antibody, clone 12C11

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€145.00
100 μG
€366.00

€145.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
25 μG
€145.00
100 μG
€366.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

€145.00


In StockDetails


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

clone

12C11, monoclonal

mol wt

calculated mol wt 170 kDa
observed mol wt ~170 kDa

purified by

using protein G

species reactivity

chicken, human, mouse

packaging

antibody small pack of 100 μg

technique(s)

electrophoretic mobility shift assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

N-terminus

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

chicken ... REST(422623)

General description

RE1-silencing transcription factor (also known as REST, Neural-restrictive silencer factor, X2 box repressor) is encoded by the REST (also known as NRSF, XBR) gene (Gene ID: 422623) in chicken. REST is a transcriptional repressor that acts by binding a DNA sequence element called the neuron-restrictive silencer element (NRSE). It is ubiquitously expressed, and higher expression is found in the tissues of the lymphocytic compartment, including spleen, thymus, peripheral blood lymphocytes, and ovary. It is also found in undifferentiated neuronal progenitor cells and it is believed that it acts as a master negative regular of neurogenesis. REST is also required to repress endogenous neuronal target genes in non-neuronal tissues, and also to repress these genes in neural precursors. Expression of REST is reported to be cell cycle-dependent with reduced levels in the G2 phase. REST restricts the expression of neuronal genes by associating with two distinct corepressors, SIN3A and RCOR1, which in turn recruit histone deacetylase to the promoters of REST-regulated genes. REST also mediates repression by recruiting the BHC complex at RE1/NRSE sites, which acts by deacetylating and demethylating specific sites on histones and acting as a chromatin modifier. REST is shown to contain nine zinc finger domains, but a C-terminally truncated form is also produced by alternative splicing. Mutations in human REST gene can lead to Wilms tumors, a pediatric malignancy of the kidney that occurs due to uncontrolled multiplication of renal stem, stromal, and epithelial cells. ((Ref.: Chen, ZF., et al. (1998). Nat. Genet. 20(2); 136-142).

Specificity

Clone 12C11 is a mouse monoclonal antibody that detects RE1-silencing transcription factor (REST). It targets an epitope within the N-terminal region.

Immunogen

GST-tagged recombinant fragment corresponding to the first 181 amino acids (without the initiator methionine) from Chicken RE1 silencing transcription factor (REST).

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from E14-Tg2a mouse embryonic stem cells.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected REST in lysate from E14-Tg2a mouse embryonic stem cells.

Tested Applications

Electrophoretic Mobility Shift Assay: A representative lot detected REST in Electrophoretic Mobility Shift Assay. (Kim, S.M., et al. (2006). Biochem Biophys Res Commun. 346(2):426-35).

Immunocytochemistry Analysis: A representative lot detected REST in Immunocytochemistry applications (Shimojo, M. et al. (2008). J Biol Chem. 283(50):34880-6).

Western Blotting Analysis: A representative lot detected REST in Western Blotting applications (Chen, Z.F., et al. (1998). Nat Genet. 20(2):136-42).

Immunoprecipitation Analysis: A representative lot immunoprecipitated REST in Immunoprecipitation applications (Shimojo, M. et al. (2008). J Biol Chem. 283(50):34880-6).

Immunohistochemistry Applications: A representative lot detected REST in Immunohistochemistry applications (Paquette, A.J., et al. (2000). Proc. Natl. Acad. Sci. USA. 97(22):12318-23).

Immunocytochemistry Analysis: A 1:25 dilution from a representative lot detected REST in NTERA-2 cell line.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-REST, clone 12C11, Cat. No. MABE1982, is a mouse monoclonal antibody that detects REST and is tested for use in Electrophoretic Mobility Shift Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service