Skip to Content
Merck
All Photos(1)

Key Documents

MABE1934

Sigma-Aldrich

Anti-IKFZ3 degron tag Antibody, clone 1C3-D3-F8

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€145.00
100 μL
€366.00

€145.00


Estimated to ship on08 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
€145.00
100 μL
€366.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€145.00


Estimated to ship on08 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C3-D3-F8, monoclonal

mol wt

calculated mol wt 58.02 kDa
observed mol wt ~39 kDa

purified by

using protein G

species reactivity

human

species reactivity (predicted by homology)

monkey, bovine, rat

packaging

antibody small pack of 100 μL

technique(s)

western blot: suitable

isotype

IgG1κ

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

General description

Zinc finger protein Aiolos (UniProt: Q9UKT9; also known as Ikaros family zinc finger protein 3, IKZF3) is encoded by the IKZF3 (also known as ZNFN1A3) gene (Gene ID: 22806) in human. IKZF3 is a member of the Ikaros Zinc finger family of transcription factors with six C2H2 type zinc finger domains. It is expressed mainly in peripheral blood leukocytes, the spleen, and the thymus. It is present as a homodimer and can also form heterodimers with other Ikaros family members. It plays an important role in the regulation of lymphocyte differentiation and is involved in the regulation of B-cell differentiation, proliferation and maturation. It is also shown to regulate Bcl-2 expression and controls apoptosis in T-cells in an IL2-dependent manner. Its levels are shown to be upregulated by TGF-1. IKZF3 expression Th17 cells is associated with increased IL-10 production and it is shown to interact with known regulators of IL10 expression. It also acts as a transcriptional activator in CD4+ T cells in cooperation with other factors, such as FOXP3 and BLIMP1 in CD4+ regulatory T cells (Tregs). The degron sequence of IKZF3 is localized to amino acids 144-168 and a Q147H mutation is reported to cripple the function of IKZF3. This sequence constitutes a modular degron that can be used to target heterologous proteins. This degron sequence is as capable as the full-length IKZF3 at targeting selected proteins for degradation. It is shown to target green fluorescent protein (GFP) for pomalidomide-induced degradation when fused to the C-terminus of GFP, but not to the N-terminus. It has also been sown to target exogenous HIF2 for pomalidomide-induced degradation when fused to the C-terminus of HIF2 . Clone 1C3-D3-F8 recognizes the degron sequence that is considered as an important regulator of protein degradation. (Ref.: Koduri, V. et al. (2019). Proc. Natl. Acad. Sci. USA. 116(7); 2539-2544).

Specificity

Clone 1C3-D3-F8 is a mouse monoclonal antibody that detects IKZF3 Protein. It targets an epitope within 25 amino acids from a Degron sequence in the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to Degron sequence from human IKZF3.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HEK293 cells expressing IKZF3 Degron-tagged PTPA.

Western Blotting Analysis (WB): A 1:500 dilution of this antibody detected Zinc finger protein Aiolos (IKZF3) in lysates from HEK293 cells ectopically expressing IKZF3 Degron-tagged PTPA, but not in lysate from untransfected HEK293 cells (wild type).
Anti-IKFZ3 degron tag, clone 1C3-D3-F8, Cat. No. MABE1934, is a mouse monoclonal antibody that detects Zinc finger protein Aiolos (IKZF3) and is tested for use in Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service