Skip to Content
Merck
All Photos(1)

Key Documents

MABE1919

Sigma-Aldrich

Anti-TNRC6ABC Antibody, clone 7A9

clone 7A9, from rat

Synonym(s):

Trinucleotide repeat-containing gene 6A, 6B, and 6C, CAG repeat protein 26, EMSY interactor protein, GW182 autoantigen, Protein GW1, Glycine-tryptophan protein of 182 kDa

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€128.00
100 μG
€350.00

€128.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€128.00
100 μG
€350.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€128.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7A9, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... TNRC6B(23112)

General description

Trinucleotide repeat-containing gene 6A, 6B, and 6C proteins (UniProt: Q8NDV7; Q9UPQ9; Q9HCJ0) are encoded by the TNRC6A, TNRC6B, and TNRC6C genes (Gene ID: 27327, 23112, and 57690), respectively in human. TNRC6 proteins are ubiquitously distributed and play a role in RNA-mediated gene silencing by both micro-RNAs (miRNAs) and short interfering RNAs (siRNAs). TNRC6 proteins are required for miRNA-dependent repression of translation and for siRNA-dependent endonucleolytic cleavage of complementary mRNAs by argonaute family proteins. They also serve as a scaffolding proteins and associate with argonaute proteins bound to partially complementary mRNAs. They can simultaneously recruit CCR4-NOT and PAN deadenylase complexes. TNRC6 proteins are predominantly found in the cytoplasm under steady state conditions, however, they are also found in the nucleus and their nuclear import is mediated via the importin pathways. TNRC6A is shown to posses both nuclear localization (NLS) as well a nuclear export signal (NES). In case of mutated NES, TNRC6A is shown to be retained in the nucleus. TNRC6 proteins contain RRM domains (aa 1781-1853; 1648-1720; and 1563-1632 for TNRC6A, 6B, and 6C, respectively). Clone 7A9 recognizes all three TNRC6 proteins. (Ref.: Schraivogel, D., et al. (2015). Nucleic Acids Res. 43(15); 7447-7461).

Specificity

Clone 7A9 is a rat monoclonal antibody that detects human Trinucleotide repeat-containing gene 6A, 6B, and 6C proteins. It target epitopes with in RRM domains.

Immunogen

Recombinant RRM domain regions from human TNRC6A, TNRC6B, and TNRC6C.

Application

Anti-TNRC6ABC, clone 7A9, Cat. No. MABE1919, is a highly specific rat monoclonal antibody that targets TNRC6 and has been tested for use in Immunofluorescence, Immunoprecipitation, and Western Blotting.
Immunofluorescence Analysis: A representative lot detected TNRC6ABC in Immunofluorescence applications (Schraivogel, D., et. al. (2015). Nucleic Acids Res. 43(15):7447-61).

Immunoprecipitation Analysis: A representative lot immunoprecipitated TNRC6ABC in Immunoprecipitation applications (Quevillon Huberdeau, M., et. al. (2017). EMBO J. 36(14):2088-2106).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in Induced Stably transfected HEK293 cell lysates.

Western Blotting Analysis: 4 µg/mL of this antibody detected TNRC6 in lysate from stably induced HEK 293 cells transfected with TNRC6A, but not in uninduced HEK 293 cells.


Target description

~175-210 kDa observed; 210.30,194.00, 175.96 kDa for TNRC6A, 6B, and 6C, respectively. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service