Skip to Content
Merck
All Photos(1)

Key Documents

MABE1526

Sigma-Aldrich

Anti-ZCCHC11 (TUT4) Antibody, clone 9F6.1

clone 9F6.1, from mouse

Synonym(s):

Terminal uridylyltransferase 4, TUTase 4, Zinc finger CCHC domain-containing protein 11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9F6.1, monoclonal

species reactivity

human, mouse

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ZCCHC11(23318)

General description

Terminal uridylyltransferase 4 (EC 2.7.7.52; UniProt Q5TAX3; also known as TUTase 4, Zinc finger CCHC domain-containing protein 11) is encoded by the ZCCHC11 (also known as KIAA0191, TUT4) gene (Gene ID 23318) in human. TUTase 4 is an uridylyltransferase that mediates the terminal uridylation of mRNAs with short (less than 25 nucleotides) poly(A) tails, hence facilitating global mRNA decay. It is involved in microRNA (miRNA)-induced gene silencing through uridylation of deadenylated miRNA targets and also acts as a suppressor of miRNA biogenesis by mediating the terminal uridylation of some miRNA precursors, including that of let-7 (pre-let-7), miR107, and miR-143 and miR-200c. It is reported to catalyze the 3′ uridylation of miR-26A that targets IL6 transcript to abrogate the silencing of IL6 transcript and promote cytokine expression. It may also participate in toll-receptor signaling by suppressing TRAF6-dependent activation of NF-kappaB.

Specificity

Clone 9F6.1 targets an epitope in the N-terminal region of ZCCHC11 (TUT4).

Immunogen

GST-tagged recombinant human ZCCHC11 (TUT4) N-terminal fragment.

Application

Anti-ZCCHC11 (TUT4), clone 9F6.1, Cat. No. MABE1526, is a highly specific mouse monoclonal antibody that targets ZCCHC11 (TUT4) and has been tested in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: 4 µg/mL from a representative lot detected ZCCHC11 (TUT4) in 10 µg of C2C12 mouse myoblast and Jurkat human T-cell lysates.

Quality

Evaluated by Western Blotting in PC3 cell lysate.

Western Blotting Analysis: 4 µg/mL of this antibody detected ZCCHC11 (TUT4) in 10 µg of PC3 human prostate cancer cell lysate.

Target description

~185 kDa observed. 185.2/81.14 kDa (human isoform 1/2) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service