Skip to Content
Merck
All Photos(1)

Key Documents

MABE1142

Sigma-Aldrich

Anti-ZP-2 Antibody, clone IE-3

clone IE-3, from rat

Synonym(s):

Zona pellucida sperm-binding protein 2, Zona pellucida glycoprotein 2, Zona pellucida protein A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€280.00

€280.00


Estimated to ship on27 May 2025Details



Select a Size

Change View
100 μG
€280.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€280.00


Estimated to ship on27 May 2025Details


biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

IE-3, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Zp2(22787)

General description

Zona pellucida sperm-binding protein 2 (UniProt: P20239; also known as Zona pellucida glycoprotein 2, Zp-2, Zona pellucida protein A) is encoded by the Zp2 (also known as Zp-2, Zpa) gene (Gene ID: 22787) in murine species. The zona pellucida is an extracellular matrix that surrounds the oocyte and early embryo. It is composed of three glycoproteins with various functions during fertilization and preimplantation development. The mammalian zona pellucida mediates species-specific sperm binding, induction of the acrosome reaction, and prevents post-fertilization polyspermy. ZP2 is expressed during the 2-week growth phase of oogenesis, prior to ovulation and may act as a secondary sperm receptor and is proteolytically cleaved in the N-terminal part by the metalloendopeptidase ASTL and is exocytosed from cortical granules after fertilization, yielding an N-terminal peptide of about 30 kDa which remains covalently attached to the C-terminal peptide via disulfide bond(s). This cleavage plays an important role in the post-fertilization block to polyspermy. Female mice lacking Zp2 gene do not form a stable zona matrix and are sterile. (Ref.: Hinsch, KD and Hinsch, E. (1999). Andrologia.31(5):320-322.)

Specificity

Clone IE-3 detects Zona pellucida sperm-binding protein 2 in murine species.

Immunogen

Epitope: N-terminus
Purified mouse zonae pellucidae.

Application

Anti-ZP-2, clone IE-3, Cat. No. MABE1142, is a highly specific rat monoclonal antibody that targets Zona pellucida sperm-binding protein 2 and has been tested in ELISA, Immunofluorescence, Immunoprecipitation, and Western Blotting.
Immunoprecipitation Analysis: A representative lot detected ZP-2 in Immunoprecipitation applications (East, I.J., et. al. (1984). J Cell Biol. 98(3):795-800).

Immunofluorescence Analysis: A representative lot detected ZP-2 in Immunofluorescence applications (East, I.J., et. al. (1984). J Cell Biol. 98(3):795-800).

Immunofluorescence A representative lot detected ZP-2 in Immunofluorescence applications (Avella, M.A., et. al. (2016). Sci Transl Med. 8(336):336ra60).

ELISA Analysis: A representative lot detected ZP-2 in Western Blotting applications (East, I.J., et. al. (1984). J Cell Biol. 98(3):795-800; Sun, W., et. al. (1999). Biol Reprod. 60(4):900-7).

Western Blotting Analysis: A representative lot detected ZP-2 in ELISA applications (Sun, W., et. al. (1999). Biol Reprod. 60(4):900-7).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in mouse ovary tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected ZP-2 in 10 µg of mouse ovary tissue lysate.

Target description

~120 kDa observed; 80.21 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service