Skip to Content
Merck
All Photos(2)

Key Documents

MABC961M

Sigma-Aldrich

Anti-RTN4IP1 Antibody, clone NIMP‐R14

clone NIMP‐R14, from rat

Synonym(s):

Reticulon-4-interacting protein 1, mitochondrial, NOGO-interacting mitochondrial protein

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€353.00

€353.00


Estimated to ship on07 May 2025Details


Request a Bulk Order

Select a Size

Change View
200 μL
€353.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€353.00


Estimated to ship on07 May 2025Details


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

NIMP‐R14, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
inhibition assay: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Reticulon-4-interacting protein 1, mitochondrial (UniProt: Q924D0; also known as NOGO-interacting mitochondrial protein, RTN4IP1) is encoded by the Rtn4ip1 (also known as Nimp) gene in murine species. RTN4IP1 is an outer mitochondrial membrane protein that interacts with reticulon 4, which is a potent inhibitor of regeneration following spinal cord injury. This interaction may be important for reticulon-induced inhibition of neurite growth. RNT4IP1 plays a role in the regulation of retinal ganglion cell (RGC) neurite outgrowth, and hence in the development of the inner retina and optic nerve. It is widely expressed in mitochondria-enriched tissues and co-localizes with the endoplasmic reticulum HSPA5 at spots corresponding to contacts with mitochondria. Mutations in Rtn4ip1 gene can cause optic atrophy 10 that leads to vision loss sometime in early childhood.

Specificity

Clone NIMP‐R14 is a rat monoclonal antibody that detects Reticulon-4-interacting protein 1 in murine species.

Immunogen

Purified BALB/c mouse neutrophils.

Application

Anti-RTN4IP1, clone NIMP‐R14, Cat. No. MABC961, is a rat monoclonal antibody that detects Reticulon-4-interacting protein 1, mitochondrial and has been tested for use in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), and Inhibition assays.
Flow Cytometry Analysis: A representative lot detected RTN4IP1 in Flow Cytometry applications (Lopez, A.F., et. al. (1984). Br J Haematol. 57(3):489-94; Taylor, P.R., et. al. (2014). Nat Immunol. 15(2):143-51).

Inhibition Analysis: A representative lot detected RTN4IP1 in Inhibition applications (Tacchini-Cottier, F., et. al. (2000). J Immunol. 165(5):2628-36; de Vries, B., et. al. (2003). J Immunol. 170(7):3883-9; Chang, H.R., et. al. (1993). J Leukoc Biol. 53(6):636-9; Mentzel, S., et. al. (1999). Kidney Int. 55(4):1335-47).

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected RTN4IP1 in mouse lung tissue.

Immunohistochemistry Analysis: A representative lot detected RTN4IP1 in Immunohistochemistry applications (Lopez, A.F., et. al. (1984). Br J Haematol. 57(3):489-94).

Immunohistochemistry Analysis: A representative lot detected RTN4IP1 in Immunohistochemistry applications (Lopez, A.F., et. al. (1984). Br J Haematol. 57(3):489-94.; de Vries, B., et. al. (2003). J Immunol. 170(7):3883-9; Chang, H.R., et. al. (1993). J Leukoc Biol. 53(6):636-9).

Immunocytochemistry Analysis: A representative lot detected RTN4IP1 in Immunocytochemistry applications (Lopez, A.F., et. al. (1984). Br J Haematol. 57(3):489-94.; Taylor, P.R., et. al. (2014). Nat Immunol. 15(2):143-51).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Immunohistochemistry in mouse spleen tissue.

Immunohistochemistry Analysis: A 1:1,000 dilution of this antibody detected RTN4IP1 in mouse spleen tissue.

Target description

43.37 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2b in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service