Skip to Content
Merck
All Photos(1)

Key Documents

MABC958

Sigma-Aldrich

Anti-TIM4/TIMD-4 Antibody, clone Kat5-18

clone Kat5-18, from hamster(Armenian)

Synonym(s):

T-cell immunoglobulin and mucin domain-containing protein 4, TIMD-4, Spleen, mucin-containing, knockout of lymphotoxin protein, SMUCKLER, T-cell immunoglobulin mucin receptor 4, TIM-4, T-cell membrane protein 4, TIM4/TIMD-4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€458.00

€458.00


Estimated to ship on30 July 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€458.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€458.00


Estimated to ship on30 July 2025Details


Request a Bulk Order

biological source

hamster (Armenian)

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Kat5-18, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
neutralization: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Timd4(276891)

General description

T-cell immunoglobulin and mucin domain-containing protein 4 (UniProt Q6U7R4; also known as Spleen mucin-containing knockout of lymphotoxin protein, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin mucin receptor 4, T-cell membrane protein 4, TIMD-4, Smuckler) is encoded by the Timd4 (also known as B430010N18Rik, Tim4) gene (Gene ID 276891) in murine species. Phagocytes, including macrophages, target apoptotic cells for engulfment by recognizing their surface exposed phosphatidylserine (PtdSer or PS). Macrophages employ specific receptors and opsonins for PS recognition, such as Milk-fat globule epidermal growth factor 8 (MFG-E8), protein S, growth arrest-specific 6 (Gas6), and TAM family TKR (Tyro 3, Axl, and MerTK) that function as protein S and Gs6 receptors. Tim (T-cell immunoglobulin and mucin domain-containing) family proteins, stabilins, and BAI1 also directly bind PtdSer and enhance the engulfment of apoptotic cells by phagocytes. Tim4 is shown to mediate murine resident peritoneal macrophages (rpMacs) engagment of apoptotic cells, while MerTK-mediates the engulfment of tethered target cells. Tim4- or MerTK-null mutations prevent rpMac-mediated apoptotic cell engulfment. Tim4-null, but not MerTK-null, macrophages lose their ability to tether apoptotic cells. Murine Tim4 is initially produced with a signal peptide sequence (a.a. 1-22), the removal of which yields the mature Tim4 with a large extracellular region (a.a.23-279), a transmembrane domain (a.a. 280-300), and a short cytoplasmic tail (a.a. 301-343).

Immunogen

Epitope: Extracellular domain
Mouse peritoneal cells.

Application

Flow Cytometry Analysis: A representative lot detected Tim4 immunoreactivity among 61% of the resident peritoneal Tim4+ MerTK+ macrophages from C57BL/6J mice (Nishi, C., et al. (2014). Mol. Cell. Biol. 34(8):1512-1520).
Flow Cytometry Analysis: Representative lots were conjugated with biotin and detected Tim4 immunoreactivity among mouse Mac1+ peritoneal cells (Miyanishi, M., et al. (2012). Int. Immunol. 24(9):551-559; Miyanishi, M., et al. (2007). Nature. 450(7168):435-439).
Neutralizing Analysis: A representative lot blocked Tim4-mediated engulfment of apoptotic CAD-/- thymocytes by murine peritoneal macrophages in a dose-dependent manner in culture (Miyanishi, M., et al. (2007). Nature. 450(7168):435-439).
Neutralizing Analysis: A representative lot, when administered via i.v. injection, significantly suppressed the phagocytosis activity of F40/80+ macrophages in the thymus of CAD-/- mice following intraperitoneal dexamethasone injection to induce apoptosis in the thymus (Miyanishi, M., et al. (2007). Nature. 450(7168):435-439).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-TIM4/TIMD-4 Antibody, clone Kat5-18 is validated for use in Flow Cytometry and Neutralizing for the detection of TIM4/TIMD-4.

Quality

Evaluated by Flow Cytometry in Ba/F3-Tim4 cells overexpressing mouse Tim4.

Flow Cytometry Analysis: 0.1 µg of this antibody detected TIM4/TIMD-4 in Ba/F3-Tim4 cells overexpressing mouse Tim4.

Target description

~37 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified armenian hamster monoclonal IgG antibody in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service