Skip to Content
Merck
All Photos(2)

Key Documents

MABC288

Sigma-Aldrich

Anti-Folliculin (FLCN), Antibody clone 8D9.2

clone 8D9.2, from mouse

Synonym(s):

Folliculin, BHD skin lesion fibrofolliculoma protein, Birt-Hogg-Dube syndrome protein, Folliculin (FLCN)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€379.00

€379.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€379.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€379.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8D9.2, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FLCN(201163)

General description

Folliculin (FLCN) is a protein with many possible roles. Folliculin may play a role in the pathogenesis of kidney cancer through its association with an inherited disorder of the hair follicle (fibrofolliculomas). But, Folliculin (FLCN) may be a tumor suppressor, however it is also involved in the generation of colorectal tumorigenesis and renal cell carcinoma, and finally Folliculin (FLCN) might be involved in energy and/or nutrient sensing through the AMPK and mTOR signaling pathways. Mutations of Folliculin (FLCN) give rise to Birt-Hogg-Dube syndrome, a rare autosomal dominant genodermatosis characterized by hair follicle hamartomas (fibrofolliculomas), kidney tumors, and spontaneous pneumothorax disorders. Folliculin (FLCN) is widely expressed and localized in both the cytoplasm and nucleus depending upon the conditions in both the fetus and the adult.

Immunogen

GST-tagged recombinant protein corresponding to human Folliculin (FLCN).

Application

Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected Folliculin (FLCN) in HeLa cells.
Alexa Fluor is a registered trademark of Life Technologies.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Folliculin (FLCN), Antibody clone 8D9.2 is validated for use in Western Blotting and Immunocytochemistry for the detection of Folliculin (FLCN).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Folliculin (FLCN) in 10 µg of HeLa cell lysate.

Target description

~65 kDa observed.

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service