Skip to Content
Merck
All Photos(2)

Key Documents

MABC287

Sigma-Aldrich

Anti-PXR Antibody, clone 1F10.1

clone 1F10.1, from mouse

Synonym(s):

Nuclear receptor subfamily 1 group I member 2, Orphan nuclear receptor PAR1, Orphan nuclear receptor PXR, Pregnane X receptor, Steroid and xenobiotic receptor, SXR, PXR

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€421.00

€421.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€421.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€421.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F10.1, monoclonal

species reactivity

human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), gorilla (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NR1I2(8856)

General description

Nuclear receptor subfamily 1 group I member 2 (UniProt O75469; also known as Orphan nuclear receptor PAR1, Orphan nuclear receptor PXR, Pregnane X nuclear receptor, Steroid and xenobiotic receptor, SXR) is encoded by the NR1I2 (also known as BXR, PAR, PRR, PXR, SAR, SXR, ONR1) gene (Gene ID 8856) in human. Pregnane X receptor (PXR) is a nuclear receptor that displays similarity to PPAR and RXR. PXR is activated by a large number of endogenous and exogenous chemicals including steroids and antibiotics. One of the key roles of PXR is the induction of CYP3A4, a P450 enzyme involved in drug metabolism.

Specificity

Epitope is present in all 7 alternative spliced isoforms (isoform 1A/PRR1-A, 1B/PRR1-B, 1C/PRR1-C, 2A/PRR2-A, 2B/PRR2-V, 2C/PRR2-C, and isoform 3).

Immunogen

KLH-conjugated linear peptide corresponding to human PXR.

Application

Detect PXR using this Anti-PXR Antibody, clone 1F10.1 validated for use in Western Blotting.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected PXR in 10 µg of human liver tissue lysate.

Quality

Evaluated by Western Blotting in human colon tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected PXR in 10 µg of human colon tissue lysate.

Target description

~53 kDa observed. Uniprot describes 7 isoforms produced by alternative splicing ranging from ~40 kDa-54 kDa

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service