Skip to Content
Merck
All Photos(3)

Key Documents

MABC189

Sigma-Aldrich

Anti-PIDD Antibody, clone 1511.1

clone 15F11.1, from mouse

Synonym(s):

p53-induced protein with a death domain, Leucine-rich repeat and death domain-containing protein, PIDD

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€421.00

€421.00


Estimated to ship on20 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€421.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€421.00


Estimated to ship on20 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

15F11.1, monoclonal

species reactivity

rat, mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PIDD1(55367)

General description

p53-induced protein with a death domain (PIDD) is also called Leucine-rich repeat and death domain-containing protein (LRDD). PIDD is involved in apoptosis downstream of the tumor suppressor. PIDD is a component of the DNA damage/stress response pathway in which p53/TP53 is associated with apoptosis. PIDD also aids in the activation of the transcription factor NF-kappa-B, and CASP2 which regulates apoptotic pathways.

Specificity

This antibody demonstrates specificity for isoform 4, but may detect isoform 1, 2 and 3. It will not detect isoforms 5, 6, and 7 based upon immunogen design.

Immunogen

Recombinant protein corresponding to human PIDD.

Application

Anti-PIDD Antibody, clone 1511.1 is a highly specific mouse monoclonal antibody, that targets p53-induced protein with a death domain & has been tested in western blotting & ICC.
Western Blotting Analysis: 1 µg/mL from a representative lot detected PIDD in 10 µg of NIH/3T3 and PC-12 cell lysate.

Immunohistochemistry Analysis: A 1:100 dilution, a 1:500 dilution, and a 1:1,000 dilution detected PIDD in HeLa, A431, and NIH/3T3 cells.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected PIDD in 10 µg of HeLa cell lysate.

Target description

~66 kDa observed. Uniprot describes 7 different isoforms.

Physical form

Format: Purified
Purified mouse IgG1κ supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service