Skip to Content
Merck
All Photos(1)

Key Documents

MABC1780

Sigma-Aldrich

Anti-CSF1R/c-FMS Antibody, clone AFS98

Synonym(s):

Macrophage colony-stimulating factor 1 receptor;EC:2.7.10.1;CSF-1 receptor;CSF-1-R;CSF-1R;M-CSF-R;Proto-oncogene c-Fms;CD115

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€139.00
100 μG
€351.00

€139.00


Estimated to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€139.00
100 μG
€351.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€139.00


Estimated to ship on14 April 2025Details


Request a Bulk Order

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

AFS98, monoclonal

mol wt

calculated mol wt 109.18 kDa

purified by

using protein G

species reactivity

mouse

packaging

antibody small pack of 100 μg

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Csf1r(12978)

General description

Macrophage colony-stimulating factor 1 receptor (UniProt: P09581; also known as EC:2.7.10.1, CSF-1 receptor, CSF-1-R, CSF-1R, M-CSF-R, Proto-oncogene c-Fms, CD115) is encoded by the Csf1r (also known as Csfmr, Fms) gene (Gene ID: 12978) in murine species. CSF1R is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 20-515), a transmembrane domain (aa 516-536), and a cytoplasmic domain (aa 537-977). It is a protein tyrosine kinase that acts as a cell surface receptor for CSF1 and IL-34. It plays an essential role in regulating cell survival, proliferation, and differentiation of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes. It promotes the release of proinflammatory chemokines in response to IL-34 and CSF1, and thereby plays an important role in innate immunity and in inflammatory processes. CSF1R is phosphorylated at multiple tyrosine residues. It is autophosphorylated in response to CSF1 or IL34 binding. Its phosphorylation at Tyr-559 is shown to be important for normal down-regulation of signaling by ubiquitination, internalization, and degradation. Its phosphorylation at Tyr-559 and Tyr-807 is important for interaction with SRC family members, including FYN, YES1 and SRC, and for subsequent activation of these protein kinases. Phosphorylation at Tyr-697 and Tyr-921 is reported to be important for its interaction with Grb2 and its phosphorylation at Tyr-721 is important for interaction with phosphoinositide-3-Kinase Regulatory Subunit 1(PIK3R1. (Ref.: Takeshita, S., et al. (2007). J. Biol. Chem. 282(26); 18980-18990; Sudo, T., et al. (1995). Oncogene. 11(12); 2469-2476; Van der Geer, P., and Hunter, T. (1993), EMBO J. 12(13); 5161-5172).

Specificity

Clone AFS98 is a rat monoclonal antibody that detects murine CSF1R/c-FMS. It targets an epitope within the extracellular domain.

Immunogen

Recombinant fragment corresponding to the extracellular domain of mouse CSF1R/c-FMS.

Application

Quality Control Testing

Evaluated by Flow Cytometry in J774.2 cells.

Flow Cytometry Analysis: 1 μg of this antibody detected CSF1R/c-FMS in one million J774.2 cells.

Tested Applications

Immunoprecipitation Analysis: A representative lot detected immunoprecipitated CSF1R/c-FMS in Immunoprecipitation applications (Sudo, T., et al. (1995). Oncogene. 11(12):2469-76).

Inhibition Assay: A representative lot inhibited colony formation of the bone marrow cells of C57BL/6 mice in response to M-CSF. (Sudo, T., et al. (1995). Oncogene.;11(12):2469-76).

Flow Cytometry Analysis: A representative lot detected CSF1R in Flow Cytometry applications (Sudo, T., et al. (1995). Oncogene. 11(12):2469-76).

ELISA Analysis: A representative lot detected CSF1R in ELISA applications (Sudo, T., et al. (1995). Oncogene. 11(12):2469-76).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-CSF1R/c-FMS, clone AFS98, Cat. No. MABC1780, is a rat monoclonal antibody that detects CSF1R/c-FMS and is tested for use in ELISA, Flow Cytometry, Inhibition Assay, and Immunoprecipitation.

Physical form

Purified rat monoclonal antibody IgG2a in PBS without azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service