Skip to Content
Merck
All Photos(2)

Key Documents

MABC1690M

Sigma-Aldrich

Anti-RAINBOW

from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG1κ

target post-translational modification

unmodified

General description

Anti-RAINBOW Antibody, Cat. No. MABC1690-25UL, validated for use in Western Blotting.
Pre-stained proteins are commonly used as molecular weight markers in electrophoresis. Several protein markers are available as pre-stained proteins with vinyl sulfone dyes. After separation through electrophoresis, the protein of interest is visualized by specific antibody coupled to a fluorophore or by chemiluminescence detection system. Anti-RAINBOW antibody is a unique antibody that can detect pre-stained molecular markers from different vendors without the need to perform a manual marking and estimating molecular weights. Anti-RAINBOW antibody detects all marker proteins in commercially available RAINBOW markers tested and lacks cross-reactivity with unstained proteins in whole cell lysates of a variety of species. It displays equal detection of Remazol-dye stained proteins: phosphorylase b (red), BSA and ADH (blue), chymotrypsin (yellow) and lysozyme (orange).

Specificity

Anti-RAINBOW is a mouse monoclonal antibody that can be used as a universal tool for visualizing prestained protein molecular weight markers in Western blot analysis. It recognizes and cross-reacts with comparable affinity with four different Remazol dye immunogens.

Immunogen

Remazol-dyes phosphorylase b (red), BSA and ADH (blue), chymotrypsin (yellow) and lysozyme (orange).

Application

Anti-RAINBOW, Cat. No. MABC1690, is a mouse monoclonal antibody that recognizes four different Remazol dye-conjugated proteins and can be used to visualize prestained molecular weight markers in Western Blotting.
Western Blotting Analysis: A representative lot detected Remazol-dye stained protein markers in Western Blotting applications (Schuchner, S., et. al. (2016). Sci Rep. 6:31363).

Western Blotting Analysis: A 1:250 and 1:500 dilution from a representative lot detected Remazol-dye Pre-Stained protein standards.

Quality

Evaluated by Western Blotting in Pre-Stained protein standard. Western Blotting Analysis: A 1:1,000, 1:5,000, and 1:10,000 dilution of this antibody detected Remzaol-dye prestained protein standards .

Physical form

Format: Purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine, 150 mM NaCl, pH 7.4, with 0.05% sodium azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service