Skip to Content
Merck
All Photos(1)

Key Documents

MABC1608M

Sigma-Aldrich

Anti-Mucin-16 (CA-125) Antibody, clone 5E11

Synonym(s):

MUC-16, Ovarian cancer-related tumor marker CA125, CA-125, Ovarian carcinoma antigen CA125

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€373.00

€373.00


Estimated to ship on06 June 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€373.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€373.00


Estimated to ship on06 June 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
proximity ligation assay: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MUC16(94025)

General description

Mucin-16 (UniProt: Q8WXI7; also known as MUC-16, Ovarian cancer-related tumor marker CA125, CA-125, Ovarian carcinoma antigen CA125) is encoded by the MUC16 (also known as CA125) gene (Gene ID: 94025) in human. Mucin-16 is a heavily O-glycosylated protein that can be liberated into the extracellular space following the phosphorylation of the intracellular C-terminus, which induces proteolytic cleavage and liberation of extracellular domain. I provides protective, lubricating barrier against particles and infectious agents at mucosal surfaces. Mucin-16 is composed of three domains: a Serine, Threonine-rich N-terminal domain, a repeated domain containing between 12 and 60 partially conserved tandem repeats of 156 amino acids and a C-terminal transmembrane domain with a short cytoplasmic tail. Its expression is up-regulated in ovarian cancer cells and may serve as a marker for ovarian cancer. Mucin-16 is shown to binds to mesothelin (MSLN) and this binding mediates heterotypic cell adhesion, which may contribute to the metastasis of ovarian cancer to the peritoneum by initiating cell attachment to the mesothelial epithelium.

Quality

Evaluated by Immunohistochemistry in human ovarian cancer and human uterine tissues.

Immunohistochemistry (Paraffin) Analysis: A 1:50-250 dilution of this antibody detected Mucin-16 in human ovarian cancer and human uterine tissues.

Target description

1519.17 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service