Skip to Content
Merck
All Photos(1)

Key Documents

MABC1227

Sigma-Aldrich

Anti-PIP4K2B (PIP5K2B) Antibody, clone 7D10.1

clone 7D10.1, from mouse

Synonym(s):

Phosphatidylinositol 5-phosphate 4-kinase type-2 beta, 1-phosphatidylinositol 5-phosphate 4-kinase 2-beta, Diphosphoinositide kinase 2-beta, Phosphatidylinositol 5-phosphate 4-kinase type II beta, PI(5)P 4-kinase type II beta, PIP4KII-beta, PtdIns(5)P-4-

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7D10.1, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PIP4K2B(8396)

General description

Phosphatidylinositol 5-phosphate 4-kinase type-2 beta (EC 2.7.1.149; UniProt P78356; also known as 1-phosphatidylinositol 5-phosphate 4-kinase 2-beta, Diphosphoinositide kinase 2-beta, Phosphatidylinositol 5-phosphate 4-kinase type II beta, PI(5)P 4-kinase type II beta, PIP4KII-beta, PIP4K2B, PtdIns(5)P-4-kinase isoform 2-beta) is encoded by the PIP4K2B (also known as PIP5K2B, PIP5KIIB) gene (Gene ID 8396) in human. PIP4K2B catalyzes the conversion of 1-phosphatidyl-1D-myo-inositol 5-phosphate to 1-phosphatidyl-1D-myo-inositol 4,5-bisphosphate. It is highly expressed in brain, heart, pancreas, skeletal muscle and kidney, however, lower levels are detected in placenta, lung, and liver. Mice deficient in PIP4K2B are shown to be hypersensitive to insulin and exhibit reduced body weights compared to wild-types. Adult male mice lacking PIP4K2B have significantly less body fat, but female mice show increased sensitivity to insulin in the presence of normal adiposity. Mice lacking PIP4K2B exhibit enhanced in vivo insulin-induced activation of Akt in the skeletal muscle and liver. Ref.: Lamia, KA., et al. (2004). Mol. Cell 24, 5080-5087

Specificity

Epitope region is present in spliced isoform 1, but not in isoform 2 reported by UniProt (P78356).

Immunogen

GST-tagged recombinant fragment corresponding to a region within the C-terminal half of human PIP4K2B (PIP5K2B).

Application

Detect PIP4K2B using this mouse monoclonal Anti-PIP4K2B (PIP5K2B) Antibody, clone 7D10.1, Cat. No. MABC1227, validated for use in Western Blotting.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in L-929 mouse fibroblast lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PIP4K2B (PIP5K2B) in 10 µg of L-929 mouse fibroblast lysate.

Target description

~48 kDa observed. 47.25/47.19 kDa (human/mouse) calculated (Met 1 removed). Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service