Skip to Content
Merck
All Photos(2)

Key Documents

MABC1191

Sigma-Aldrich

Anti-Sodium-Iodide Symporter Antibody, clone ARN1

clone ARN1, from mouse

Synonym(s):

Sodium/iodide cotransporter, Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, Solute carrier family 5 member 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€341.00

€341.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€341.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€341.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ARN1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... SLC5A5(6528)

General description

Sodium/iodide cotransporter (UniProt: Q92911; also known as Na(+)/I(-) cotransporter, Sodium-iodide symporter, Na(+)/I(-) symporter, Solute carrier family 5 member 5) is encoded by the SLC5A5 (also known as NIS) gene (Gene ID: 6528) in human. Sodium/iodide cotransporter is a multi-pass membrane protein that mediates iodide uptake in thyroid gland. It is expressed primarily in thyroid tissue, but is present in lower levels in mammary gland and ovary. It is found in the basolateral membrane of thyroid follicular cells where it mediates efficient iodide uptake from the bloodstream into the thyroid using the sodium gradient generated by Na+/K+-ATPase. It transports one iodide ion along with two sodium ions following the inwardly directed sodium gradient. Sodium/iodide cotransporter expression is significantly reduced in tumors. The presence of symporter in mammary glands leads to secretion of iodine in milk that is required for thyroid function in neonatal animals. NIS gene and protein expression is stimulated by thyroid-stimulating hormone. Both human and rat NIS contain N-glycosylation sites, however, glycosylation is not required for its full activity. Mutations in SLC5A5 gene are known to cause thyroid dyshormonogenesis (TDH1) that is characterized by the inability of the thyroid to maintain a concentration difference of readily exchangeable iodine between the plasma and the thyroid gland, leading to congenital hypothyroidism. Autoantibodies to NIS adversely affect iodide transport.

Specificity

Clone ARN1 is a mouse monoclonal antibody that detects Sodium/iodide cotransporter in human cells. It targets an epitope within 13 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 13 amino acids from the C-terminal region of human Sodium/iodide cotransporter.

Application

Anti-Sodium-Iodide Symporter, clone ARN1, Cat. No. MABC1191, is a mouse monoclonal antibody that detects Sodium/iodide cotransporter and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected Sodium-Iodide Symporter in human stomach tissue sections.

Immunohistochemistry (Paraffin) Analysis: A representative lot detected Sodium-Iodide Symporter in Immunohistochemistry applications (Tazebay, U.H., et. al. (2000). 6(8):871-8).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in MDCK cells transduced with human WT NIS.

Western Blotting Analysis: 1 µg/mL of this antibody detected Sodium-Iodide Symporter in MDCK cells transduced with human WT Sodium-Iodide Symporter (NIS).

Target description

~68 kDa observed; 68.67 kda calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 140 mM Tris and 180 mM Glycine (pH 7.4) without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service