Skip to Content
Merck
All Photos(1)

Key Documents

MAB984

Sigma-Aldrich

Anti-Yellow Fever Virus Antibody, clone 2D12.A

ascites fluid, clone 2D12.A, Chemicon®

Synonym(s):

Yellow fever

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€337.00

€337.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€337.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€337.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

clone

2D12.A, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

inhibition assay: suitable (hemagglutination)
neutralization: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Reacts with the envelope protein of the wild (Asibi) and vaccine strains of yellow fever virus. Preferentially neutralizes the Asibi strain. No cross-reactivity with other flaviviruses has been demonstrated.

Immunogen

17D strain of yellow virus.

Application

Neutralization (BLK) Asibi YF; 1:1000-1:3000.

Hemagglutination-inhibition: 1:3000-1:6000.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Yellow Fever Virus Antibody, clone 2D12.A is validated for use in HI, NEUT for the detection of Yellow Fever Virus.

Physical form

Ascites.

Storage and Stability

Maintain at -20°C for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yellow Fever-induced Acute Lung Injury.
Alexandre Todorovic Fabro et al.
American journal of respiratory and critical care medicine, 200(2), 250-252 (2019-02-26)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service