Skip to Content
Merck
All Photos(1)

Documents

MAB8599

Sigma-Aldrich

Anti-RSV Antibody, fusion protein, all type A, B strains, clone 131-2A

clone 131-2A, Chemicon®, from mouse

Synonym(s):

RSV Fusion

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

131-2A, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG2aκ

shipped in

wet ice

General description

RSV is a labile paramyxovirus that produces a characteristic fusion of human cells in tissue culture--the syncytial effect. Two subtypes, A and B, have been identified. Subtype B are characterized as the asymptomatic strains of the virus. The more severe clinical illnesses involve Subtype A strains.

Specificity

Reacts with RSV fusion protein, specifically epitope F1a of RSV F-protein {Sominina AA, et al Vestn Ross Akad Med Nauk. 1995;(9):49-54} found on all A & B strains of RSV.

Immunogen

A2 RSV virus cell extract
Epitope: all type A & B strains

Application

Anti-RSV Antibody, fusion protein, all type A, B strains, clone 131-2A detects level of Respiratory Syncytial Virus & has been published & validated for use in ELISA, FC, IF & WB.
Indirect immunofluorescence.

ELISA

Immunoblotting: (using purified F protein preparations, clone identified a ~70kDa band, Tripp, RA et al 2003)

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Target description

~70 kDa

Physical form

Format: Purified
Protein A Purified immunoglobulin in 0.02M phosphate, 0.25M NaCl, 0.1% NaN3 pH 7.6
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
RSV Control Slides, Catalogue Number 5012-5

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Tripp et al.
Journal of virology, 73(9), 7099-7107 (1999-08-10)
BALB/c mice sensitized to vaccinia virus expressed G protein of respiratory syncytial virus (RSV) develop a Th2-type cytokine response and pulmonary eosinophilia when challenged with live RSV. In this study, BALB/c mice were immunized or challenged with an RSV mutant
Subgrouping of respiratory syncytial virus strains from Australia and Papua New Guinea by biological and antigenic characteristics
Hierholzer, J C, et al
Archives of Virology, 136, 133-147 (1994)
Real-time detection of virus particles and viral protein expression with two-color nanoparticle probes
Agrawal, Amit, et al
Journal of virology, 79, 8625-8628 (2005)
Anti-G protein antibody responses to respiratory syncytial virus infection or vaccination are associated with inhibition of G protein CX3C-CX3CR1 binding and leukocyte chemotaxis
Harcourt, Jennifer L, et al
The Journal of Infectious Diseases, 190, 1936-1940 (2004)
Ivy Widjaja et al.
PloS one, 10(6), e0130829-e0130829 (2015-06-25)
The respiratory syncytial virus (RSV) fusion protein F is considered an attractive vaccine candidate especially in its prefusion conformation. We studied whether recombinant soluble RSV F proteins could be stabilized in a prefusion-like conformation by mutation of heptad repeat B

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service