Skip to Content
Merck
All Photos(1)

Key Documents

MAB844-3

Sigma-Aldrich

Anti-Parainfluenza 2 Blend Antibody, clones 231/9F, 233/9A

Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€288.00

€288.00


Estimated to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€288.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€288.00


Estimated to ship on07 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

clone

231/9F, monoclonal
233/9A, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable

isotype

IgG

shipped in

dry ice

Specificity

Specific for the HN protein common to all parainfluenza II strains. No cross-reactivity to Parainfluenza I, III or IV. Blend of MAB844-1 (clone 231/9F) and MAB844-2 (clone 233/9A).

Immunogen

Greer

Application

Anti-Parainfluenza 2 Blend Antibody, clones 231/9F, 233/9A detects level of Parainfluenza 2 Blend & has been published & validated for use in ELISA & IF.
ELISA: 1:800+.

Indirect Immunofluorescence: 1:100-200+ (fresh frozen tissue sections).

Optimal working dilutions must be determined by end user.



Not suggested for use in immunoprecipitation.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Ascites fluid
Format: Purified

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service