Skip to Content
Merck
All Photos(1)

Documents

MAB8184

Sigma-Aldrich

Anti-EBV VCA gp125 Antibody, clone L2

clone L2, Chemicon®, from mouse

Synonym(s):

VCA-gp125

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

L2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

Specificity

VCA-gp125

Application

Anti-EBV VCA gp125 Antibody, clone L2 is an antibody against EBV VCA gp125 for use in ELISA, IF & WB.
Immunofluorescence

ELISA

Immunoblotting (weak)

Optimal dilutions must be determined by the end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
Purified immunoglobulin. In 0.02M phosphate buffer, 0.25M sodium chloride, pH= 7.6 with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katerina Vrzalikova et al.
Blood, 117(22), 5907-5917 (2011-03-18)
An important pathogenic event in Epstein-Barr virus (EBV)-associated lymphomas is the suppression of virus replication, which would otherwise lead to cell death. Because virus replication in B cells is intimately linked to their differentiation toward plasma cells, we asked whether
Glycoprotein gp110 of Epstein-Barr virus determines viral tropism and efficiency of infection.
Neuhierl, B; Feederle, R; Hammerschmidt, W; Delecluse, HJ
Proceedings of the National Academy of Sciences of the USA null
Primary B-cell infection with a deltaBALF4 Epstein-Barr virus comes to a halt in the endosomal compartment yet still elicits a potent CD4-positive cytotoxic T-cell response.
Bernhard Neuhierl,Regina Feederle,Dinesh Adhikary,Birgit Hub,Karsten Geletneky et al.
Journal of virology null
Julien Batisse et al.
Journal of virology, 79(22), 14102-14111 (2005-10-29)
Most human herpesviruses, including Epstein-Barr virus (EBV), express a protein which functions primarily as an mRNA export factor. Previously, we deleted the gene for the Epstein-Barr virus mRNA export factor EB2 from the EBV genome and then introduced the mutated
Hongda Huang et al.
Nature communications, 7, 12707-12707 (2016-09-02)
The histone H3.3 chaperone DAXX is implicated in formation of heterochromatin and transcription silencing, especially for newly infecting DNA virus genomes entering the nucleus. Epstein-Barr virus (EBV) can efficiently establish stable latent infection as a chromatinized episome in the nucleus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service