Skip to Content
Merck
All Photos(1)

Key Documents

MAB8129

Sigma-Aldrich

Anti-Cytomegalovirus Antibody, immediate early, clone 7D12.2

clone 7D12.2, Chemicon®, from mouse

Synonym(s):

CMV

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€820.00

€820.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€820.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

€820.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

7D12.2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

suitability

not suitable for immunohistochemistry (Paraffin)

shipped in

wet ice

Specificity

Reacts with an immediate early protein M.W. 68 kD. Can detect CMV infection 1 hour post-infection exhibiting a nuclear and cytoplasmic staining which reaches peak intensity between 8-24 hours.

With CMV the antigens expressed at different times are listed as:



Immediate Early (alpha gene expression): those antigens expressed at 3-12 hrs post-infection generally involved in Transcription such as 72kD major phosphoprotein and a few other other antigens at 60 - 80kD.

Early Antigen (beta genes) a.k.a. Delayed Early or Intermediate Early: expressed at 12-24hrs post-infection. Generally enzymes and one virion structural gene preceding viral DNA synthesis.

Late Antigen (gamma genes): expressed at 36-48hrs post-infection. Generally structural proteins. Major protein = 55kD

Immunogen

Epitope: immediate early
MRC-5 cells infected with CMV AD169.

Application

Immunohistochemistry.

Immunocytochemistry.

IFA at 1:3,000-1:6,000 on acetone fixed cells.

Does not work on paraffin embedded tissue sections.

Dilute with buffer pH 7.4-7.6 to desired working volume.

For extensive dilution, protein containing or other stabilizing medium should be used.

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Cytomegalovirus Antibody, immediate early, clone 7D12.2 is validated for use in IF, WB, IC, IH for the detection of Cytomegalovirus.

Physical form

Format: Purified
Purified ascites in 0.02 M Phosphate Buffered Saline, 0.25 M Sodium Chloride, 0.1% Sodium Azide,

pH 7.6 .

Storage and Stability

Maintain at +2° to + 8°C for up to 12 months from date of receipt.

During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200 μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Shimizu et al.
Japanese journal of ophthalmology, 62(6), 667-676 (2018-09-08)
Human cytomegalovirus (HCMV) infections can cause endotheliitis which is associated with an elevation in the intraocular pressure (IOP). However, the mechanism of the IOP elevation has not been established. The purpose of this study was to determine whether HCMV strains

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service