Skip to Content
Merck
All Photos(7)

Key Documents

MAB4401

Sigma-Aldrich

Anti-Oct-4 Antibody, clone 10H11.2

clone 10H11.2, Chemicon®, from mouse

Synonym(s):

Octamer-binding transcription factor 3, POU class 5 homeobox 1, POU domain class 5, transcription factor 1, POU domain, class 5, transcription factor 1, POU-type homeodomain-containing DNA-binding protein, octamer-binding transcription factor-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

clone

10H11.2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

input

sample type induced pluripotent stem cell(s)
sample type: human embryonic stem cell(s)

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POU5F1(5460)

General description

Octamer-4 (Oct-4), a member of the POU family of transcription factors, has been demonstrated to be vital for the formation of self-renewing pluripotent stem cells. During embryogenesis expression of Oct-4 is limited to pluripotent cells of the inner cell mass (ICM) that contribute to the formation of all fetal cell types. This relationship between Oct-4 and pluripotency has seen this transcription factor emerge as a marker of pluripotent stem cells. Undifferentiated human and murine pluripotent Embryonic Stem (ES) and Embryonic Carcinoma (EC) cells express Oct-4. Additionally, murine Embryonic Germ (EG) cells are also known to express Oct-4. Following stem cell differentiation, the level of Oct-4 expression decreases. Oct-4 expression by human EG cells is not known.
Purified monoclonal antibody for the detection of Human Oct-4.

Specificity

Reacts with human Oct-4 only, and detects the Oct-4A isoform. (Note: Please order Catalog No. MAB4419 for use with mouse embryonic stem cells).
The clone reacts strongly with specific nuclear binding to human ES cells and cross-reacts weakly with mouse ES cells.

Application

Detect Oct-4 using this Anti-Oct-4 Antibody, clone 10H11.2 validated for use in WB, FC, IC, ELISA.
Flow Cytometry: A previous lot of this antibody was used in FC.

ELISA: A previous lot of this antibody was used in ELISA.

Immunocytochemistry: A previous lot of this antibody was used in IC. H9 cells in culture on a mouse embronic fibroblast feeder layer were labeled by a standard indirect IF protocol. Oct-4 labelling in green, DAPI in blue, & SSEA-3 (MAB4303 in red).

Optimal working dilutions must be determined by the end user.

Quality

Evaluated by western blot on mouse P1 brain lysates.

Western Blotting: 1:500 dilution of this antibody detected Oct-4 on 10 µg of mouse P1 brain lysates

Target description

44 kDa

Physical form

Format: Purified

Analysis Note

Control
Human embryonic stem cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microporous membrane growth substrates for embryonic stem cell culture and differentiation.
Steven D Sheridan,Sonia Gil,Matthew Wilgo,Aldo Pitt
Methods in Cell Biology null
Efficient derivation of embryonic stem cells from nuclear transfer and parthenogenetic embryos derived from cryopreserved oocytes.
Sung LY, Chang CC, Amano T, Lin CJ, Amano M, Treaster SB, Xu J, Chang WF, Nagy ZP, Yang X, Tian XC
Cellular Reprogramming null
Lingjun Rao et al.
Nature communications, 9(1), 126-126 (2018-01-11)
The generation of functional skeletal muscle tissues from human pluripotent stem cells (hPSCs) has not been reported. Here, we derive induced myogenic progenitor cells (iMPCs) via transient overexpression of Pax7 in paraxial mesoderm cells differentiated from hPSCs. In 2D culture
Chen Liu et al.
PloS one, 9(9), e108239-e108239 (2014-09-27)
Annulus fibrosus (AF) injuries can lead to substantial deterioration of intervertebral disc (IVD) which characterizes degenerative disc disease (DDD). However, treatments for AF repair/regeneration remain challenging due to the intrinsic heterogeneity of AF tissue at cellular, biochemical, and biomechanical levels.
Li Zou et al.
Scientific reports, 6, 22868-22868 (2016-03-17)
Human pluripotent stem cells provide a standardized resource for bone repair. However, criteria to determine which exogenous cells best heal orthopedic injuries remain poorly defined. We evaluated osteogenic progenitor cells derived from both human embryonic stem cells (hESCs) and induced

Articles

Skip weekend feedings. Defined serum-free and feeder-free expansion media for human pluripotent stem cells (ES and iPS cells). See publications and protocols.

Fibroblast growth factors (FGFs) regulate developmental pathways and mesoderm/ectoderm patterning in early embryonic development.

The Simplicon™ RNA Reprogramming Technology is a next generation reprogramming system that uses a single synthetic, polycistronic self-replicating RNA strand engineered to mimic cellular RNA to generate human iPS cells.

Human iPSC neural differentiation media and protocols used to generate neural stem cells, neurons and glial cell types.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service