Skip to Content
Merck
All Photos(1)

Key Documents

MAB4356A4

Sigma-Aldrich

Anti-DPPA-2 Antibody, clone 6C1.2, Alexa Fluor 488 conjugate

clone 6C1.2, from mouse, ALEXA FLUOR 488

Synonym(s):

Pluripotent embryonic stem cell-related gene 1 protein, developmental pluripotency associated 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€382.00

€382.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€382.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€382.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C1.2, monoclonal

species reactivity

mouse

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Dppa2(73703)

General description

DPPA-2 (developmental pluripotency associated-2) is a novel Oct-4-related protein that is localized to the nucleus of mouse fetal germ cells. DPPA-2 has binding sites for both POU and Sox-2 protein domains and is expressed primarily in the developing germ line following gastrulation. These associations suggest that this protein participates in a very strong supportive role in maintaining cell pluripotentiality. In addition to its role as a pluripotency marker, more recent research using cancer cells suggests that DPPA-2 may also act as an embryo-cancer antigen—pointing to the presence of abnormal cell growth and the formation of cancerous cells.

Immunogen

recombinant protein corresponding to mouse DPPA-2.

Application

Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation
This Anti-DPPA-2 Antibody, clone 6C1.2, Alexa Fluor 488 conjugate is validated for use in ICC for the detection of DPPA-2.

Quality

Evaluated by Immunocytochemistry in Mouse Embryonic Stem Cells (SCR012). Immunocytochemsitry Analysis: A 1:100 dilution of this antibody detected SOX-2 in Mouse Embryonic Stem Cells (SCR012).

Target description

~30 kDa observed

Physical form

Protein G Purified
Purified mouse monoclonal IgG2aλ supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8ºC protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Mouse Embryonic Stem Cells (SCR012).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service