Skip to Content
Merck
All Photos(2)

Key Documents

MAB4234-C

Sigma-Aldrich

Anti-Wilms′ Tumor Antibody, NT clone 6F-H2, Ascites Free

clone 6F-H2, from mouse

Synonym(s):

Wilms tumor protein, WT33

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€388.00

€388.00


Estimated to ship on22 April 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1677


Select a Size

Change View
100 μG
€388.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€388.00


Estimated to ship on22 April 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1677

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6F-H2, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WT1(7490)

General description

Wilms tumor protein (UniProt P19544; also known as WT33) is encoded by the WT1 (also known as DDS, FS, MEACHS, MESOM, NPHS4, WAGR) gene (Gene ID 7490) in human. The Wilms’ tumor gene WT1 was originally identified in the childhood kidney cancer Wilms’ tumor. The N-terminal region of WT1 protein contains a proline-rich region (a.a. 27-83) involved in transcriptional regulation, self-association, and RNA recognition, while its C-terminal region contains four zinc fingers (a.a 323-347, 353-377, 383-405, 414-438) that mediate DNA and RNA binding. The zinc finger domain of WT1 can bind to GC-rich sequences, such as the EGR-1 consensus sequence (5’-GCG(T/G)GGGCG-3’), the WTE motif (5′-GCGTGGGAGT-3′), or (TCC)n motif. Many genes responsible for cell growth and apoptosis, such as Bcl-2, Bcl-xL, BFL1, and c-myc, have been identified as downstream targets of WT1. There are four major alternatively spliced WT1 isoforms resulting from splicing at either or both of exon 5 (17AA) and exon 9 (KTS). All four major WT1 isoforms are overexpressed in leukemia and solid tumors and play oncogenic roles such as inhibition of apoptosis, and promotion of cell proliferation, migration and invasion.

Application

Immunohistochemistry Analysis: An 1:250 dilution from a representative lot detected Wilms tumor protein in human kidney tissue.

Immunoprecipitation Analysis: A representative lot co-immunoprecipitated CRE-binding protein/CBP together with Wilms tumor protein WT1 from the lysate of a T-SV40 immortalized human glomerular epithelial cell (HGEC) line (Drossopoulou, G.I., et al. (2009). Am. J. Physiol. Renal Physiol. 297(3):F594-F603).

Western Blotting Analysis: A representative lot detected Wilms tumor protein WT1 in the CRE-binding protein/CBP immunoprecipitate obtained from the lysate of a T-SV40 immortalized human glomerular epithelial cell (HGEC) line (Drossopoulou, G.I., et al. (2009). Am. J. Physiol. Renal Physiol. 297(3):F594-F603).

Western Blotting Analysis: A representative lot detected Wilms tumor protein WT1 in lysates from mouse E15.5 embryonic kidney and human melanoma cell lines A375, SK-MEL-28, and WM-266-4 (Wagner, N., et al. (2008). Pflugers Arch. Eur. J. Physiol. 455(5):839-847).

Immunocytochemistry Analysis: A representative lot immunostained the nucleus of methanol-fixed human melanoma A375 cells by fluorescent immunocytochemistry (Wagner, N., et al. (2008). Pflugers Arch. Eur. J. Physiol. 455(5):839-847).

Immunofluorescence Analysis: A representative lot immunostained the PCNA-positive nuclei of proliferating cells in formalin-fixed, paraffin-embedded human melanoma tissue sections by fluorescent immunohistochemistry (Wagner, N., et al. (2008). Pflugers Arch. Eur. J. Physiol. 455(5):839-847).

Immunohistochemistry Analysis: A representative lot immunostained glomeruli in formalin-fixed, paraffin-embedded normal human kidney and Wilms′ tumor sections (Wagner, N., et al. (2008). Pediatr. Nephrol. 23(9):1445-1453).

Immunohistochemistry Analysis: A representative lot detected vascular WT1 expression in 95% of 113 paraffin-embedded tumour tissues of various types. In most cases, nuclear WT1 staining of endothelial cells was seen (Wagner, N., et al. (2008). Oncogene. 27(26):3662-3672).

Immunohistochemistry Analysis: A representative lot immunostained the nucleus of perifollicular fibroblasts at the hair follicle in formalin-fixed, paraffin-embedded normal human skin sections. Most common melanocytic nevi do not express WT1, whereas Spitz nevi and dysplastic nevi show cytoplasmic WT1 staining. (Wagner, N., et al. (2008). Pflugers Arch. Eur. J. Physiol. 455(5):839-847).
This Anti-Wilms′ Tumor Antibody, NT clone 6F-H2, Ascites Free is validated for use in Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting for the detection of Wilms′ tumor protein.

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Wilms tumor protein in 10 µg of Jurkat cell lysate.

Target description

~52 kDa observed. 49.19 kDa (isoform 1), 47.20 kDa (isoform 2), 47.51 kDa (isoform 3), 48.87 kDa (isoform 4), 34.45 kDa (isoform 5), 56.88 kDa (isoform 6), 55.21 kDa (isoform 7), 33.09 kDa (isoform 8) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service