Skip to Content
Merck
All Photos(1)

Key Documents

MAB3834

Sigma-Aldrich

Anti-Polyhistidine Tag Antibody

Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€521.00

€521.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€521.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€521.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Reacts with the epitope MRGSHHHHHH which is present in pQE vectors. It is expected that the antibody will also react with proteins expressed from vectors such as PRSET A, B, and C. The antibody should not react with pAC, N, T, A, B, C baculovirus vectors.

Immunogen

His tagged recombinant protein.

Application

Detect Polyhistidine Tag using this Anti-Polyhistidine Tag Antibody validated for use in ELISA & WB.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
Western blot: 1:2,000

ELISA

Optimal working dilutions must be determined by end user.

Linkage

Replaces: MAB3112

Physical form

Format: Purified
Purified immunoglobulin. Liquid containing 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana L Valenciano et al.
Cell cycle (Georgetown, Tex.), 15(20), 2827-2841 (2016-09-03)
The Trypanosoma brucei subspecies T. brucei gambiense and T. brucei rhodesiense are vector-borne pathogens that cause sleeping sickness also known as Human African Trypanosomiasis (HAT), which is fatal if left untreated. The drugs that treat HAT are ineffective and cause
Benjamin Geary et al.
Frontiers in immunology, 14, 1290585-1290585 (2023-12-14)
MZB1 is an endoplasmic reticulum residential protein preferentially expressed in plasma cells, marginal zone and B1 B cells. Recent studies on murine B cells show that it interacts with the tail piece of IgM and IgA heavy chain and promotes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service