Skip to Content
Merck
All Photos(4)

Key Documents

MAB3786-C

Sigma-Aldrich

Anti-APC Antibody, CT, clone C-APC Antibody 28.9, Ascites Free

clone C-APC 28.9, from mouse

Synonym(s):

Adenomatous polyposis coli protein, Deleted in polyposis 2.5, Protein APC

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€317.00

€317.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€317.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€317.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C-APC 28.9, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... APC(324)

General description

Adenomatous polyposis coli protein (UniProt P25054; also known as Deleted in polyposis 2.5, Protein APC) is encoded by the APC (also known as DP2.5) gene (Gene ID 324) in human. The multifunctional tumor suppressor APC is a large scaffold protein that contains multiple armadillo repeat (ARM) domains (a.a. 453-767 of human APC spliced isoform 1) for mediating various cellular functions, including Wnt signal regulation, cell adhesion, and chromosomal segregation during mitosis. APC, AXIN, casein kinase 1 (CK1) and glycogen synthase kinase 3 (GSK3) constitute the core proteins of the β-catenin destruction complex that functions as the negative regulator against Wnt signaling transduction. Mice with Apc gene mutations develop adenomas in the intestine due to Wnt hyperactivation, and functional loss of APC due to gene mutations is the hallmark of human colorectal cancers (CRC). APC also directs cholinergic synapse assembly between neurons and is commonly employed as a marker for monitoring stem cells neuronal differentiation.

Specificity

Recognizes human APC, molecular weight is approximately 310 kDa.

Immunogen

Epitope: C-terminal region.
Human APC C-terminal fusion with MBP (maltose binding protein).

Application

Anti-APC Antibody, CT, clone C-APC Antibody 28.9, Ascites Free is an antibody against APC for use in Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected APC in human kidney and cardiac muscle.
Immunocytochemistry Analysis: A representative detected an increased APC expression in human eyelid adipose-derived stem cells (hEASCs) after neuroinduction at passage 8 (Zhou, J., et al. (2014). J. Cell. Mol. Med. 18(2):326-343).
Immunofluorescence Analysis: A representative lot detected a granular APC staining pattern at a subapical region of epithelial cells in fallopian mucosa by fluorescent immunohistochemistry staining of paraffin-embedded, paraformaldehyd-fixed fallopian tube tissue sections (Kessler, M., et al. (2012). Am. J. Pathol. 180(1):186-198).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected APC in human cerebral cortex tissue.

Target description

311.6 kDa calculated.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service