Skip to Content
Merck
All Photos(1)

Documents

MAB329

Sigma-Aldrich

Anti-Synaptophysin Antibody, clone SP15

ascites fluid, clone SP15, Chemicon®

Synonym(s):

Major Synaptic Vesicle Protein p38

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

SP15, monoclonal

species reactivity

pig, rat, hamster, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgM

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SYP(6855)

Specificity

Synaptophysin. Reacts with synaptophysin expressed in CHO cells.

Immunogen

Crude synaptic immunoprecipitate (Human)

Application

Immunohistochemistry at 1:200-1:5,000.

Formalin fixed (fixed <4 weeks), paraffin embedded

Frozen

4% Paraformaldehyde

Western blotting at 1:2,500-1:10,000.

ELISA at 1:500

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
This Anti-Synaptophysin Antibody, clone SP15 is validated for use in ELISA, WB, IH(P) for the detection of Synaptophysin.

Linkage

Replaces: 04-1019

Physical form

Liquid

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michelle A Land et al.
Scientific reports, 9(1), 16737-16737 (2019-11-15)
A non-synonymous single nucleotide polymorphism of the human serotonin 5-HT2C receptor (5-HT2CR) gene that converts a cysteine to a serine at amino acid codon 23 (Cys23Ser) appears to impact 5-HT2CR pharmacology at a cellular and systems level. We hypothesized that
Gerald D Griffin et al.
The Journal of comparative neurology, 518(22), 4531-4545 (2010-10-05)
Central oxytocin (OT) modulates many social behaviors, including female rat sexual receptivity, quantified as the copulatory stance known as lordosis. The expression of the lordosis response is modulated by OT action in the ventromedial nucleus of the hypothalamus (VMH), as
A Odawara et al.
Scientific reports, 6, 26181-26181 (2016-05-18)
The functional network of human induced pluripotent stem cell (hiPSC)-derived neurons is a potentially powerful in vitro model for evaluating disease mechanisms and drug responses. However, the culture time required for the full functional maturation of individual neurons and networks
Camille Belzil et al.
The Journal of biological chemistry, 288(34), 24452-24464 (2013-07-19)
In acute and chronic neurodegeneration, Ca(2+) mishandling and disruption of the cytoskeleton compromise neuronal integrity, yet abnormalities in the signaling roles of cytoskeletal proteins remain largely unexplored. We now report that the microtubule-associated protein p600 (also known as UBR4) promotes
Raymond S Roginski et al.
Gene, 648, 42-53 (2018-01-18)
The known functions of the human GCOM1 complex hub gene include transcription elongation and the intercalated disk of cardiac myocytes. However, in all likelihood, the gene's most interesting, and thus far least understood, roles will be found in the central

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service