Skip to Content
Merck
All Photos(1)

Key Documents

MAB1951Z

Sigma-Aldrich

Anti-Integrin beta 1 (CD29) Antibody

CHEMICON®, mouse monoclonal, P4G11

Synonym(s):

CD29, MAB1951

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€459.00

€459.00


Estimated to ship on22 April 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1230

Request a Bulk Order

Select a Size

Change View
100 μG
€459.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€459.00


Estimated to ship on22 April 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1230

Request a Bulk Order

Product Name

Anti-Integrin β1 Antibody, clone P4G11, azide free, clone P4G11, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P4G11, monoclonal

species reactivity

monkey, human, rabbit

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)

Specificity

Human integrin beta1. The epitope recognized by MAB1951Z is Ca2+ dependent.

Immunogen

Human IL-1beta activated endothelial cells

Application

Immunohistochemistry: for use on acetone or mild paraformaldehyde fixed tissue (2-2.5% PFA, short time) frozen and 4% PFA paraffin embedded {Qin et al 2003 1:500, citrate HEIR treated}

Ca++ required for binding. Ca++ must be used in blocking, washing, and antibody incubation buffers.

Traditional formalin fixation is not recommended.

Immunocytochemistry: 2 ug/mL (for use on numerous cell lines as long as Ca2+ is present)

Immunoprecipitation: 5 μg/400 μL lysate (0.5% triton PBS plus Ca++)

FACS: 2-5μg per 10e6 cells, in the presence of Ca++.

Stimulation of cells: 2-10 μg/ml per 10e6 cells. Binding buffer must contain 1mM Ca++.

ELISA

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins
This Anti-Integrin β1 Antibody, clone P4G11, azide free is validated for use in ELISA, FC, IP, FUNC, FUNC, IC, IH for the detection of Integrin β1.

Physical form

Format: Purified
Sterile-filtered protein A purified immunoglobulin in 0.01M PBS, pH 7.1, containing no preservatives.

Storage and Stability

Maintain between 2 and 8°C for up to 6 months from date of receipt. Avoid freeze thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Todd J Herron et al.
Circulation. Arrhythmia and electrophysiology, 9(4), e003638-e003638 (2016-04-14)
Human pluripotent stem cell-derived cardiomyocytes (hPSC-CMs) monolayers generated to date display an immature embryonic-like functional and structural phenotype that limits their utility for research and cardiac regeneration. In particular, the electrophysiological function of hPSC-CM monolayers and bioengineered constructs used to
Garima Dwivedi et al.
Cartilage, 12(2), 237-250 (2018-12-21)
Previous studies have shown that intrinsic behavior of subchondral bone marrow stem cells (BMSCs) is influenced by donors and locations. To understand the variability in cartilage repair outcomes following bone marrow stimulation, we tested the hypothesis that in vivo cartilage
Garima Dwivedi et al.
Cartilage, 9(4), 378-390 (2017-11-22)
Bone marrow stimulation procedures initiate repair by fracturing or drilling subchondral bone at base of cartilaginous defect. Earlier studies have shown that defect location and animal age affect cartilage repair outcome, suggesting a strong influence of structural and biological characteristics
Hsin-Ling Yin et al.
International journal of molecular sciences, 17(9) (2016-09-03)
Triple negative breast cancer (TNBC) displays higher risk of recurrence and distant metastasis. Due to absence of estrogen receptor (ER), progesterone receptor (PR) and human epidermal growth factor receptor 2 (HER2), TNBC lacks clinically established targeted therapies. Therefore, understanding of
Stephen P Evanko et al.
Matrix biology : journal of the International Society for Matrix Biology, 42, 74-92 (2015-01-01)
The contribution of hyaluronan-dependent pericellular matrix to TGF-β1-driven induction and maintenance of myofibroblasts is not understood. Hyaluronan is an extracellular matrix (ECM) glycosaminoglycan important in cell adhesion, proliferation and migration, and is implicated in myofibroblast formation and maintenance. Reduced turnover

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service