MAB19310-C
Anti-Aggrecan Antibody, MMP Cleaved, clone AF-28, Ascites Free
clone AF-28, from mouse
Synonym(s):
Aggrecan core protein, MMP-cleaved, Cartilage-specific proteoglycan core protein, MMP-cleaved, Chondroitin sulfate proteoglycan 1, MMP-cleaved, Chondroitin sulfate proteoglycan core protein 1, MMP-cleaved, CSPCP, MMP-cleaved
Select a Size
€373.00
Estimated to ship on11 February 2025Details
Select a Size
About This Item
€373.00
Estimated to ship on11 February 2025Details
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
AF-28, monoclonal
species reactivity
bovine, porcine, human, rat, mouse
technique(s)
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... ACAN(176)
General description
Specificity
Immunogen
Application
Immunohistochemistry Analysis: A representative lot detected MMP-cleaved aggrecan in paraffin-embedded tibia sections from both wild-type mice and mice with decreased MMP-13 expression (Zhou, X., et al. (2010). Proc. Natl. Acad. Sci. U. S. A. 107(29):12919-12924).
Western Blotting Analysis: A representative lot detected a higher level of MMP-generated FFGV fragments in osteoarthritis/osteoarthritic (OA) cartilage than in normal cartilage samples (Struglics, A., and Hansson, M. (2012). Biochem. J. 446(2):213-223).
Western Blotting Analysis: A representative lot detected MMPs-digested G2 fragment from recombinant G1-G2, but not undigested, elastase- or trypsin-digested fragments (Mercuri, F.A., et al. (1999). J. Biol. Chem. 274(45):32387-32395).
Western Blotting Analysis: A representative lot detected MMP-13-digested fragments from human, pig, bovine and rat, but not shark, aggrecan. Clone AF-28 did not detect undegraded aggrecan (Fosang, A.J., et al. (1996). FEBS Lett. 380(1-2):17-20).
Western Blotting Analysis: A representative lot detected in vitro generated aggrecan fragments by MMPs, as well as various aggrecan fragments in arthritis patients-derived synovial fluids (Fosang, A.J., et al. (1995). Biochem. J. 310( Pt 1):337-343).
Cell Structure
ECM Proteins
Quality
Western Blotting Analysis: 0.5 µg/mL of this antibody detected MMP-cleaved aggrecan in 10 µg of human chondrocyte tissue lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service