Skip to Content
Merck
All Photos(1)

Key Documents

MAB1615SP

Sigma-Aldrich

Anti-Neurofilament 70 kDa, clone DA2 (mouse monoclonal)

Sign Into View Organizational & Contract Pricing

Select a Size

90 μL
€200.00

€200.00


Estimated to ship on25 April 2025Details


Request a Bulk Order

Select a Size

Change View
90 μL
€200.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€200.00


Estimated to ship on25 April 2025Details


Request a Bulk Order

Quality Level

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service