Skip to Content
Merck
All Photos(1)

Key Documents

MAB1397Z

Sigma-Aldrich

Anti-ICAM-1 Antibody, clone 3E2B, Azide Free

clone 3E2B, Chemicon®, from hamster(Armenian)

Synonym(s):

CD54, MAB1397

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€421.00

€421.00


Estimated to ship on30 March 2026Details


Request a Bulk Order

Select a Size

Change View
100 μG
€421.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€421.00


Estimated to ship on30 March 2026Details


Request a Bulk Order

biological source

hamster (Armenian)

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

3E2B, monoclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Icam1(15894)

Specificity

MAB1371Z is reactive with mouse ICAM-1 (CD54), which is present on T-cells, B-cells, monocytes and endothelial cells, epithelial cells, dendritic cells and keratinocytes. Expression of CD54 is increased after stimulation with cytokines and LPS, and it also plays a role in B cell-T cell interactions.

Application

Anti-ICAM-1 Antibody, clone 3E2B, Azide Free detects level of ICAM-1 & has been published & validated for use in ELISA, FC, FUNC, IC, IH.
Immunocytochemistry: Live or acetone fixed cells

ELISA

Immunohistochemistry of acetone fixed frozen sections. (Paraffin not tested).

Flow Cytometry: Suggested working dilution 50 μg/mL. Use 5 mL of this dilution to label 0.5 x 10(e6) cells.

Immunoprecipitation: use protein G or rabbit anti-hamster IgG secondary

Clone 3E2B is reported to be function blocking against murine ICAM-1.

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Linkage

Replaces: CBL1331F

Physical form

Format: Purified
Purified IgG prepared by affinity chromatography on Protein G agarose beads. Liquid at 1 mg/mL in sterile-filtered PBS, containing no preservatives. Endotoxin level 4.13EU/mg

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze-thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dominique Promeneur et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(3), 1035-1040 (2013-01-02)
Tragically common among children in sub-Saharan Africa, cerebral malaria is characterized by rapid progression to coma and death. In this study, we used a model of cerebral malaria appearing in C57BL/6 WT mice after infection with the rodent malaria parasite

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service