Skip to Content
Merck
All Photos(1)

Key Documents

MAB10234

Sigma-Aldrich

Anti-Hepatitis B Virus Surface Antigen Antibody, clone H21F8-1

ascites fluid, clone H21F8-1, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€369.00

€369.00


Estimated to ship on08 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€369.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€369.00


Estimated to ship on08 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

H21F8-1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgM

shipped in

wet ice

General description

Hepatitis B virus surface antigen is one of the earliest markers of infection with hepatitis B virus. Hepatitis B virus surface antigen is tested for in the differential diagnosis of hepatitis. It is the first detectable viral antigen to appear during infection with this virus; however, early in an infection, this antigen may not be present and it may be undetectable later in the infection as it is being cleared by the host.

Application

Anti-Hepatitis B Virus Surface Antigen Antibody, clone H21F8-1 detects level of Hepatitis B Virus Surface Antigen & has been published & validated for use in IF.

Storage and Stability

Stable for 12 months when stored at -20°C from date of shipment

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinical Characteristics and Surgical Outcome in Hepatocellular Carcinoma without Hepatitis B Virus Surface Antigen or Hepatitis C Virus Antibody
Tanaka, Kuniya, et al
Annals of Surgical Oncology, [Epub ahead of print]-[Epub ahead of print] (2006)
Sang Hyun Nam et al.
Archives of pharmacal research, 29(11), 1042-1048 (2006-12-07)
Plasmid DNA vaccines encoding the hepatitis B virus (HBV) surface and hepatitis C virus (HCV) envelope antigens, respectively, were constructed, and attempt were made to find the possibility of a divalent vaccine against HBV and HCV. The expression of each

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service