Skip to Content
Merck
All Photos(1)

Key Documents

MAB10210

Sigma-Aldrich

Anti-Disialosyl LEA Antigen Antibody, clone FH7

ascites fluid, clone FH7, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€539.00

€539.00


Estimated to ship on09 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€539.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€539.00


Estimated to ship on09 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

FH7, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG3

shipped in

wet ice

target post-translational modification

unmodified

General description

The disialosyl Lea antigen is found in various cancer tissues and their cell lines such as colonic and gastric cancers. It is absent in various normal tissues and blood cells. Sera of patients with early cases of colonic and gastric cancers, display an elevated disialosyl Lea antigen level,

Immunogen

Purified Disialosyl Lea antigen

Application

Anti-Disialosyl LEA Antigen Antibody, clone FH7 detects level of Disialosyl LEA Antigen & has been published & validated for use in IF.
Research Category
Apoptosis & Cancer
Research Sub Category
Tumor Markers

Physical form

Ascites
Liquid ascites fluid

Storage and Stability

1 year at 4°C from date of shipment

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service