Skip to Content
Merck
All Photos(2)

Key Documents

LUXRGTSTK

Millipore

mPAGE® Lux Bis-Tris Stacking Solution for mPAGE® Lux Curing Station

greener alternative

Stacking Solution for MPAGE® Lux Curing Station

Synonym(s):

lux bis-tris stacking solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€67.20

€67.20


Estimated to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EACH
€67.20

About This Item

UNSPSC Code:
41106514
NACRES:
NC.01

€67.20


Estimated to ship on03 April 2025Details


Request a Bulk Order

Product Name

mPAGE® Lux Bis-Tris Stacking Solution for mPAGE® Lux Curing Station,

description

Reagent for use with mPAGE® Lux Curing Station

Quality Level

packaging

1 ea of 1 ea

manufacturer/tradename

Millipore

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

electrophoresis: suitable
immunoblotting: suitable
western blot: suitable

greener alternative category

General description

Use the mPAGE® Lux Bis-Tris Stacking Solution with mPAGE® Lux Curing Station, mPAGE® Lux Resolving Solution, and mPAGE® Lux Diluent for making polyacrylamide gels. It offers rapid and simple gel curing process for handcasting mini protein gels. A mini gel is cured in 1.5min and overall casting process can be less than 3min.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Carc. 1B - Eye Dam. 1 - Muta. 1B - Repr. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Questions

  1. Is mPAGE Lux gel compatible with BIORAD tank?

    1 answer
    1. The mPAGE Lux solutions are designed for use with the Lux curing station. The gels are compatible with the BIORAD mini protean tanks. However, the BIORAD casters do not fit within the mPAGE Lux casting station.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service