Skip to Content
Merck
All Photos(1)

Documents

LSKMAGN04

Millipore

PureProteome NHS FlexiBind Magnetic Bead System

NHS FlexiBind beads are suitable for Immunoprecipitations, purifying nucleic acids, isolating cells and organelles, performing protein-protein interaction studies and many other applications.

Synonym(s):

Magnetic Bead System, NHS FlexiBind Bead System

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
eCl@ss:
32160405
NACRES:
NA.56

form

liquid

packaging

pkg of 2 mL

manufacturer/tradename

PureProteome

technique(s)

protein purification: suitable

particle size

10 μm

capacity

>17 μmol/mL, settled beads binding capacity (NHS)

shipped in

wet ice

storage temp.

2-8°C

General description

{TM="PureProteome"] NHS (N-Hydroxysuccinimide) FlexiBind Magnetic Beads provide researchers flexibility in binding the ligand of their choice. The only prerequisite is that the molecule must contain a primary free amine group. NHS FlexiBind beads are being used for the generation of custom affinity beads for binding to target ligands and protein purification.

Application

PureProteome NHS FlexiBind Magnetic Bead System has been used in co-immunoprecipitation and affinity purification.

Legal Information

PureProteome is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ling Liu et al.
European journal of pharmacology, 858, 172520-172520 (2019-07-07)
The metabolic disorder of succinate in myocardial tissue during ischemia-reperfusion can lead to the myocardial oxidative injury. The activation of succinate dehydrogenase (SDH) plays a vital role in the process. Silent information regulator 5 (Sirt5), a nicotinamide adenine dinucleotide (NAD)-dependent
Zakhar O Shenkarev et al.
Journal of neurochemistry, 155(1), 45-61 (2020-03-31)
Lynx1 is a GPI-tethered protein colocalized with nicotinic acetylcholine receptors (nAChRs) in the brain areas important for learning and memory. Previously, we demonstrated that at low micromolar concentrations the water-soluble Lynx1 variant lacking GPI-anchor (ws-Lynx1) acts on α7-nAChRs as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service