Skip to Content
Merck
All Photos(1)

Key Documents

LSKMAGD12

Millipore

PureProteome Albumin/IgG Depletion Kit

The PureProteome Albumin/IgG Magnetic Beads are conjugated to an antibody specific for human serum albumin and to protein G for capturing IgG.

Synonym(s):

IgG Removal Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€1,270.00

€1,270.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
€1,270.00

About This Item

UNSPSC Code:
41116133
eCl@ss:
32160405
NACRES:
NA.56

€1,270.00


Please contact Customer Service for Availability

packaging

kit of 12 mL beads

manufacturer/tradename

PureProteome

technique(s)

depletion: suitable (serum)
protein purification: suitable

particle size

10 μm

shipped in

wet ice

storage temp.

2-8°C

General description

The PureProteome Albumin/IgG Magnetic Beads are conjugated to an antibody specific for human serum albumin and to protein G for capturing IgG. These magnetic beads provide a rapid, scalable, and reproducible means to deplete >98% of both albumin and IgG from serum and plasma samples, facilitating the detection and analysis of proteins of interest. For efficient depletion, the PureProteome Magnetic Beads must be used with the PureProteome Magnetic Stand.

Components

PureProteome Albumin/IgG Magnetic Beads, 12ml

10X Phosphate Buffered Saline (PBS) wash and bind buffer, 7ml

Amicon Ultra-4 3K Centrifugal Filter, 8pk

Analysis Note

Depletion: >98% depletion of Albumin and IgG

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rongrong Ren et al.
Experimental and therapeutic medicine, 19(6), 3627-3633 (2020-04-30)
The incidence of acute lung injury (ALI) and acute respiratory distress syndrome (ARDS) is a common health problem in the clinic and is projected to increase in prevalence in the future. Mechanical ventilation is commonly used to provide respiratory support

Questions

  1. Does LSKMAGD12 have compatibility with both mouse and rat?

    1 answer
    1. The PureProteome albumin and albumin/IgG beads removed over 85% of the albumin in mouse serum and over 75% of the albumin in rat serum.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service