LA1HCG6S2
Millistak+® Lenticular Disc Filter
A1HC, cellulose, filtration area 1.8 m2, hydrophilic
Synonym(s):
Millistak+ Lenticular Disc Filters, A1HC media series, 16 in. diameter, 8-cell, silicone seal, 2/pack
About This Item
Recommended Products
material
cellulose
filter aid (inorganic)
mixed cellulose esters (MCE)
polypropylene
silicone seal
Quality Level
description
Post-TFF (Prostak™) clarification fluids
feature
hydrophilic
manufacturer/tradename
Millistak+®
parameter
0.03 bar max. differential pressure (0.5 psid) (Reverse)
0.2 bar max. inlet pressure (3 psi) at 123 °C
0.21 bar max. differential pressure (3 psid) at 123 °C (Forward)
1 bar max. inlet pressure (15 psi) at 80 °C
1.03 bar max. differential pressure (15 psid) at 80 °C (Forward)
2.07 bar max. differential pressure (30 psid) at 25 °C (Forward)
2.1 bar max. inlet pressure (30 psi) at 25 °C
diam.
40.64 cm (16 in.)
filtration area
1.8 m2
impurities
<0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)
non-volatile residue (NVR)
1.50-2.02 mg/m2 (gravimetric analysis test)
total organic carbon (TOC) residue
416-593 ppb (gravimetric analysis test)
565-1037 ppb
input
sample type liquid
conductivity
5-7 μmho per cm at 25 °C (product water)
filter grade
A1HC
Looking for similar products? Visit Product Comparison Guide
General description
Application
Specifications
Preparation Note
May be hot water sanitized for 20 cycles using purified water @ 80 °C for 30 minutes
May be steam sterilized for 3 steam-in-place (SIP) cycles of 30 minutes @ 123 °C
Analysis Note
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT RE 1 Inhalation
Target Organs
Lungs
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service