Skip to Content
Merck
All Photos(10)

Key Documents

IX70XXPKIT

Milli-Q

Milli-Q® IX 7010/15 Water Purification Kit

greener alternative

For Milli-Q® IX 7010/15 pure water systems. Protects RO membranes and stored pure water quality. This product replaces IX70XXPKT1.

Sign Into View Organizational & Contract Pricing

Select a Size

IX70XXPKIT
€1,650.00

€1,650.00


Available to ship on11 April 2025Details


Request more information

Select a Size

Change View
IX70XXPKIT
€1,650.00

About This Item

UNSPSC Code:
41104212

€1,650.00


Available to ship on11 April 2025Details


Request more information

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

Milli-Q®

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

humidity measuring range

10-80% (In original sealed packaging to protect purification elements)

H

39.5 cm (15.5 in.)

input

feed water nature potable tap water (within specifications of the water purification system)

compatibility

for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015

greener alternative category

shipped in

ambient

storage temp.

2-30°C

General description

This water purification kit includes:

  • Two IPAK Gard® pretreatment cartridges composed of a pleated filter for the removal of particulates, minerals and colloids and a carbon block for the removal of free chlorine. This protects the reverse osmosis (RO) membrane and improves system performance.
  • The tank vent filter protects stored pure (Type 2) water from airborne contaminants. It contains activated carbon, soda lime and a membrane filter to prevent impure air from entering the tank.

We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Application

  • For use with Milli-Q® IX 7010/15 pure water systems
  • IPAK Gard® pretreatment cartridges protect RO membranes to ensure system performance over years
  • Tank vent filter protects stored pure water from airborne contaminants

Features and Benefits

  • e-Sure tags enable RFID connection with the system, for seamless consumable status monitoring, data management and full traceability
  • Supplied with one Drain Cap per cartridge to remove excess water and reduce the weight of exhausted the cartridges at disposal
  • 63% of the parts suppliers for the IPAK Gard® cartridge are ISO 14001 environmental management system-certified

Legal Information

IPAK GARD is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Reduce your lab’s consumption of water, energy, plastics and chemicals. Eco-friendly Milli-Q® lab water purification systems and digital services can support your lab’s sustainability goals.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service