Skip to Content
Merck
All Photos(1)

Key Documents

HTS206M

Sigma-Aldrich

ChemiSCREEN Membrane Preparation Recombinant Human MrgD Mas-Related Gene Family Receptor

Human MRGPRD / MrgD GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.

Sign Into View Organizational & Contract Pricing

Select a Size

200 UNITS
€685.00

€685.00


Estimated to ship on02 May 2025Details



Select a Size

Change View
200 UNITS
€685.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

€685.00


Estimated to ship on02 May 2025Details


biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Full-length human MRGPRD cDNA encoding MrgD
The Mas-related genes (Mrgs), also known as sensory neuron-specific G-protein coupled receptors (SNSRs), are selectively expressed in subpopulations of sensory neurons involved in the perceptor of pain (Dong et al., 2001). The Mrg family in rodents consists of 50 receptors in the MrgA-MrgH subfamilies, but humans contain fewer Mrg members. MrgD (also known as TGR7) is found across species from rodents to humans, and its expression is restricted to nonpeptidergic, small-diameter, IB4+ dorsal root ganglion (DRG) neurons. MrgD binds β-alanine, but not L-alanine, which results in increased intracellular calcium and the suppression of forskolin-induced cAMP production, thus suggesting coupling to Gq and Gi proteins (Shinohara et al., 2004). β-alanine-induced activation of MrgD inhibits KCNQ2/3-mediated M-currents in cells overexpressing MrgD and KCNQ2/3 and also in cultured DRG neurons, which indicates that MrgD enhances nociceptive signaling (Crozier et al., 2007). Millipore′s MrgD membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of MrgD interactions with its ligands. The cell line exhibits a calcium response with EC50s of 11 µM for β-alanine. The membrane preparations exhibit EC50s of 69 µM for β-alanine in a GTPS binding assay.

Application

Radioligand binding assay and GTPγS binding.

Biochem/physiol Actions

GPCR Class: A
Protein Target: MRGPRD / MrgD
Target Sub-Family: Mas-related gene

Quality

SPECIFICATIONS: 1 unit = 5 µg membrane preparation
EC50 in GTPγS binding assay by β-alanine: ~ 69 μM

Specifications

Inucbation Conditions
ASSAY CONDITIONS: Membranes are permeabilized by addition of saponin to an equal concentration by mass, then mixed with [35S]-GTPγS (final concentration of 0.3 nM) in 20 mM HEPES, pH 7.4/100 mM NaCl/10 mM MgCl2/0.5 μM GDP in a nonbinding 96-well plate. Unlabeled β-alanine was added to the final concentration indicated in Figure 1 (final volume 100 μL), and incubated for 30 min at 30°C. The binding reaction is transferred to a GF/B filter plate (Millipore MAHF B1H) previously prewetted with water. The plate is washed 3 times (1 mL per well per wash) with cold 10 mM sodium phosphate, pH 7.4, then dried and counted.

One vial contains enough membranes for at least 200 assays (units), where one unit is the amount of membrane that will yield greater than 1000 cpm specific β-alanine -stimulated [35S]-GTPγS binding.

The MrgD membrane preparation is expected to be functional in a radioligand binding assay; however, the end user will need to determine the optimal radiolabeled ligand for use with this product.

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA no preservatives.
Packaging method: Membranes protein were adjusted to 1 mg/ml in 1 ml packaging buffer, rapidly frozen, and stored at -80°C

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Dong et al.
Cell, 106(5), 619-632 (2001-09-12)
In vertebrates, peripheral chemosensory neurons express large families of G protein-coupled receptors (GPCRs), reflecting the diversity and specificity of stimuli they detect. However, somatosensory neurons, which respond to chemical, thermal, or mechanical stimuli, are more broadly tuned. Here we describe
Robert A Crozier et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 27(16), 4492-4496 (2007-04-20)
The recently identified Mas-related gene (Mrg) family of G-protein-coupled receptors is expressed almost exclusively in dorsal root ganglion (DRG) neurons. The expression of one family member, MrgD, is even further confined to IB4+, nonpeptidergic, small-diameter nociceptors. Although the functional consequences
Tokuyuki Shinohara et al.
The Journal of biological chemistry, 279(22), 23559-23564 (2004-03-24)
We isolated a cDNA encoding an orphan G protein-coupled receptor, TGR7, which has been recently reported to correspond to MrgD. To search for ligands for TGR7, we screened a series of small molecule compounds by detecting the Ca2+ influx in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service