Skip to Content
Merck
All Photos(1)

Documents

HTS020RTA

Sigma-Aldrich

Ready-to-Assay CB2 Cannabinoid Receptor Frozen Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76

description

Cell line type: GPCR Cell Lines
GPCR Class A
protein target CB2

Quality Level

technique(s)

calcium flux assay: suitable

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

Cannabinoid compounds include exogenous drugs such as 9-THC, the main psychoactive component of the plant Cannabis sativa, and endogenous mediators, such as anandamide, that belong to eicosanoid family. The biological effects of cannabinoids are mediated by a family of two Gi-coupled 7-transmembrane receptors, CB1 and CB2. The CB1 receptor is found primarily in brain and mediates the psychoactive effects of cannabinoid ligands. The CB2 receptor is expressed mainly in immune cells, including mast cells and CD40-activated B cells, where it mediates proliferation and inhibition of migration (Howlett et al., 2002). Activation of CB2 inhibits the development of liver fibrosis (Julien et al., 2005). In bone, CB2 is expressed in both osteoblasts and osteoclasts, and functions to prevent bone loss (Ofek et al., 2006). In addition, activation of CB2 has an antinociceptive effect in animal models of neuropathic, inflammatory, and acute pain; this effect is mediated by release of endogenous opioids in the periphery (Ibrahim et al., 2005). Millipore′s cloned human CB2-expressing cell line is made in the Chem-4 host, which supports high levels of recombinant CB2 expression on the cell surface and contains optimized levels of a recombinant promiscuous G protein to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists, and modulators at CB2.

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial. Fifty (50) mL of Media Component.

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service