Skip to Content
Merck
All Photos(1)

Key Documents

HATF29325

Millipore

Immobilon-NC Membrane, Triton-free, mixed cellulose esters, 0.45 µm, 293 mm, disc

0.45 um ( 0.45 micron filter ) pore size Mixed Cellulose Esters Transfer Membrane for western blotting using Chromogenic, Radioactive or Fluorescent detection methods.

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
€539.00

€539.00


Available to ship on15 April 2025Details



Select a Size

Change View
25 EA
€539.00

About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602

€539.00


Available to ship on15 April 2025Details


material

mixed cellulose esters (MCE) (surfactant-free)
plain white filter

Quality Level

feature

hydrophilic

manufacturer/tradename

Immobilon®

technique(s)

western blot: suitable

filter diam.

293 mm

thickness

180 μm

refractive index

n/D 1.51

pore size

0.45 μm pore size

capacity

117 μg/cm2 (insulin)
160 μg/cm2 adsorption capacity (BSA)
259 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with Amido black
for use with CPTS
for use with Colloidal gold
for use with India ink
for use with Ponceau-S red

detection method

colorimetric
fluorometric
radioactive

General description

Filter Code: HATF
Primary Binding Mechanisms: Electrostatic, hydrophobic

Application

0.45 um ( 0.45 micron filter ) pore size Mixed Cellulose Esters Transfer Membrane for western blotting using Chromogenic, Radioactive or Fluorescent detection methods.

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: BioProcessing
  • Intended Use: Reduction or removal of microorganism/bioburden
  • Instructions for Use: Please refer first page of Visual Inspection Guide
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service