Skip to Content
Merck
All Photos(1)

Key Documents

HATF08250

Millipore

Immobilon®-NC Membrane

82 mm disc, 0.45 µm pore size, Triton-free, mixed cellulose esters

Synonym(s):

Mixed cellulose ester membrane

Sign Into View Organizational & Contract Pricing

Select a Size

50 PKG
€211.00

€211.00


Available to ship on14 April 2025Details



Select a Size

Change View
50 PKG
€211.00

About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.22

€211.00


Available to ship on14 April 2025Details


Product Name

Immobilon®-NC Membrane, Triton-free, mixed cellulose esters, 0.45 m, 82 mm, disc, 0.45 um ( 0.45 micron filter ) pore size Mixed Cellulose Esters, Surfactant-Free, Transfer Membrane for western blotting using Chromogenic, Radioactive or Fluorescent detection methods.

material

mixed cellulose esters (MCE) membrane (surfactant-free)
nitrocellulose membrane
plain filter
white filter

Quality Level

feature

hydrophilic

manufacturer/tradename

Immobilon®

technique(s)

western blot: suitable

filter diam.

82 mm

thickness

180 μm

refractive index

n/D 1.51

pore size

0.45 μm pore size

capacity

117 μg/cm2 adsorption capacity (insulin)
160 μg/cm2 adsorption capacity (BSA)
259 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with Amido black
for use with CPTS
for use with Colloidal gold
for use with India ink
for use with Ponceau-S red

detection method

chemiluminescent
colorimetric
fluorometric
radioactive

shipped in

ambient

General description

Filter Code: HATF
Primary Binding Mechanisms: Electrostatic, hydrophobic

Application

  • Biological solutions
  • Cell contact
  • Very small volumes requiring surfactant-free surfaces
  • Isolation of virus-like particles in wastewater
  • Microplastics analysis grade water
  • Nucleic acid binding, including eDNA

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service