GTBP04700
Polycarbonate Membrane Filter, 0.22 μm Pore Size
Isopore™, filter diam. 47 mm, hydrophilic, brown
Synonym(s):
Isopore Membrane Filter
Select a Size
€426.00
Available to ship on15 April 2025Details
Select a Size
About This Item
€426.00
Available to ship on15 April 2025Details
Recommended Products
material
brown filter
plain filter
polycarbonate membrane
sterility
non-sterile
feature
hydrophilic
manufacturer/tradename
Isopore™
Millipore
sustainability
Greener Alternative Product
parameter
140 °C max. temp.
6 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi)
~16 mL/min-cm2 water flow rate at 10 psi (typical results)
diam.
47 mm
filter diam.
47 mm
thickness
25 μm
gravimetric extractables
<1%
color
brown
refractive index
n/D 1.6
matrix
Isopore™
pore size
0.22 μm pore size
5-20 % porosity
bubble point
≥5.2 bar, air with water at 23 °C
greener alternative category
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Application
- Epifluorescent microscopy
- Particle monitoring
- Air monitoring
- Microplastics analysis
- Microplastics analysis grade water
Caution
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service