Skip to Content
Merck
All Photos(1)

Key Documents

GSS-1016

Sigma-Aldrich

GS System GS Media Supplement 50X, Mixture of non-essential amino acids & nucleosides for addition to glutamine-free basal media

The GS System (offered by & a registered trademark of Celltech Biologicals, Inc.) utilizes specific vectors for cell culture transfection.

Synonym(s):

50X GS Supplement, GS Media Supplement, Non-Essential Amino Acids

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€118.00

€118.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€118.00

About This Item

UNSPSC Code:
12352205
eCl@ss:
32160801
NACRES:
NA.75

€118.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

Quality Level

form

liquid

manufacturer/tradename

Specialty Media

General description

The GS System (offered by and a registered trademark of Celltech Biologicals, Inc.) utilizes specific vectors for cell culture transfection. Specialty Media offers media and reagents for use with this system. Our GS System products are made using the highest quality reagents and our high purity Ultra Pure H2O.

Storage and Stability

Store at -20°C upto 1 year.

Legal Information

GS System is a trademark of Lonza Group Ltd.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service