Skip to Content
Merck
All Photos(1)

Key Documents

GR32L

Sigma-Aldrich

Anti-Glucocorticoid Receptor (Ab-2) Mouse mAb (BuGR2)

lyophilized, clone BuGR2, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€553.00

€553.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€553.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€553.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

BuGR2, monoclonal

form

lyophilized

contains

≤0.1% sodium azide as preservative

species reactivity

guinea pig, rabbit, rat, mouse, sheep

should not react with

amphibian, primate, avian

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG2a

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Nr3C1(14815)

General description

Anti-Glucocorticoid Receptor (Ab-2), mouse monoclonal, clone BuGR2, recognizes ~94-97 kDa glucocorticoid receptor in mouse liver. It is validated for WB, ICC, IP & IHC (Frozen & paraffin sections).
Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with P3-AgX-653 myeloma cells (see application references). Recognizes the ~94-97 kDa glucocorticoid receptor.
Recognizes the ~94-97 kDa glucocorticoid receptor in mouse liver tissue.

Immunogen

Rat
partially purified, rat glucocorticoid receptor

Application


Frozen Sections (5 g/ml)
Immunoblotting (5 g/ml)
Immunocytochemistry (5 g/ml)
Immunoprecipitation (see comments)
Paraffin Sections (5 g/ml)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from PBS, pH 7.2.

Reconstitution

Resuspend the lyophilized antibody with 0.1 ml sterile PBS, pH 7.2. Be careful to reconstitute the entire contents of the vial; during shipment and handling, portions of the lyophilized pellet may have become dislodged and may not be in the bottom of the vial. Following reconstitution, aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solutions.

Analysis Note

Positive Control
mouse liver extract

Other Notes

Does not cross react with androgen, progesterone, estrogen or mineralocorticoid receptors. Enzymatic digestion and immunoblot analysis has shown that this antibody reacts with the undigested 97 kDa glucocorticoid receptor, a 17 kDa DNA-binding trypsin fragment, and a 45 kDa steroid and DNA-binding chymotrypsin fragment. Immunocytochemical localization of the glucocorticoid receptor is observed in the cytoplasm and nucleus, even in the presence of hormone. This antibody has also been reported to work for immunoprecipitation. Antibody should be titrated for optimal results in individual systems.
Hussien, A., et al. 1997. Proc. Natl. Acad. Sci. USA94, 1521.
Pratt, W.B. and Toft, D.O., 1997. Endocrine Rev.18 306.
Schumacher, M., et al. 1996. Dev. Neurosci.18, 6.
Toran-Allerand, C.D. 1996. Dev. Neurosci.18, 36.
Weigel, N.L. 1996. Biochem J.319, 657.
Jones, K.J. 1994. Ann. N.Y. Acad. Sci.743, 141.
Arnold, A.P. 1990. in Hormones, Brain and Behavior in Vertebrates, ed. Balthazart, J. (Karger, Basel), 82.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad M Khan et al.
Neuroendocrinology, 84(5), 317-329 (2006-12-05)
MNAR/PELP1 (see text) is a newly identified scaffold protein/coactivator initially thought to modulate nongenomic and genomic actions of the estrogen receptor; however, it has been recently shown to interact with multiple steroid receptors, including androgen and glucocorticoid receptors. In the
Biswajyoti Sahu et al.
Cancer research, 73(5), 1570-1580 (2012-12-28)
The forkhead protein FoxA1 has functions other than a pioneer factor, in that its depletion brings about a significant redistribution in the androgen receptor (AR) and glucocorticoid receptor (GR) cistromes. In this study, we found a novel function for FoxA1
Nina M Patrick et al.
The Journal of steroid biochemistry and molecular biology, 167, 1-13 (2016-09-21)
Small molecule inhibitors of lysine deacetylases (KDACs) are approved for clinical use in treatment of several diseases. Nuclear receptors, such as the glucocorticoid receptor (GR) use lysine acetyltransferases (KATs or HATs) and KDACs to regulate transcription through acetylation and deacetylation
Vineela Kadiyala et al.
The Journal of biological chemistry, 288(40), 28900-28912 (2013-08-16)
Nuclear receptors use lysine acetyltransferases and lysine deacetylases (KDACs) in regulating transcription through histone acetylation. Lysine acetyltransferases interact with steroid receptors upon binding of an agonist and are recruited to target genes. KDACs have been shown to interact with steroid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service