Skip to Content
Merck
All Photos(1)

Key Documents

GR15L

Sigma-Aldrich

Anti-EGFR (Ab-5) Mouse mAb (EGFR.1)

lyophilized, clone EGFR.1, Calbiochem®

Synonym(s):

Anti-Epidermal Growth Factor Receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€614.00

€614.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€614.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€614.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

EGFR.1, monoclonal

form

lyophilized

does not contain

preservative

species reactivity

human

should not react with

mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG2b

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... EGFR(1956)
mouse ... Egfr(13649)
rat ... Egf(25313)

General description

Anti-EGFR (Ab-5), mouse monoclonal, clone EGFR.1, recognizes the ~170 kDa EGF receptor protein in A431 cells. It is validated for use in FC, IF, IP, and neutralization studies.
Purified mouse monoclonal antibody generated by immunizing mice with the specified immunogen and fusing splenocytes with NS-1 mouse myeloma cells. Recognizes the ~170 kDa EGF receptor protein.
Recognizes the ~170 kDa EGF receptor protein in A431 cells.

Immunogen

Human
partially purified, human EGFR from A431 cells

Application


Flow Cytometry (1-5 g/ml)
Immunofluorescence (1-5 g/ml)
Immunoprecipitation (1-2 g/sample)
Neutralization Studies (1-2 g/ml; see application references)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from a volatile buffer, 100 µg BSA.

Reconstitution

We recommend resuspending the lyophilized antibody with sterile PBS, pH 7.4, or sterile 20 mM Tris-saline (20 mM Tris containing 0.15 M NaCl), pH 7.4, to yield a final concentration of 100 µg/ml; product will be more stable if 0.1% sodium azide is included (do not add azide if antibody is to be used with viable cells). Lyophilized antibodies should be resuspended at 4°C with occasional gentle mixing for at least 2 h.

Analysis Note

Negative Control
HL-60 cells
Positive Control
A-431 cells

Other Notes

Hunter T., 1984. Nature311, 414.
Reynolds, F.H., Jr., et al. 1981. Nature292, 259.
Ushiro, H. and Cohen, S. 1980. J. Biol. Chem.255, 8363.
Stains A431, HeLa-5 cells and other human cells but not to rodent cells. It inhibits EGF binding to its receptor and is an antagonist of in vivo EGF-stimulated tyrosine kinase activity. This antibody does not cross-react to c-neu/erb B-2. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Duan et al.
Asian Pacific journal of cancer prevention : APJCP, 13(9), 4255-4261 (2012-11-22)
The present study was conducted to evaluate radioimmunoimaging (RII) and in vivo distribution of mixed antibodies 99mTc-EGFR-mAb and 99mTc-CD44- mAb in nude mice bearing human lung adenocarcinoma xenografts. Single and mixed applications of the two radiolabeled monoclonal antibodies (mAbs) were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service