Skip to Content
Merck
All Photos(1)

Key Documents

GF442-M

Sigma-Aldrich

HumanKine® TNF alpha, Human Recombinant Xeno-Free

Synonym(s):

Tumor Necrosis Factor-α, TNFa

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€5,230.00

€5,230.00


Estimated to ship on20 May 2025Details



Select a Size

Change View
1 MG
€5,230.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

€5,230.00


Estimated to ship on20 May 2025Details


Assay

>95% (SDS-PAGE)

Quality Level

impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

UniProt accession no.

shipped in

dry ice

General description

Tumor necrosis factor alpha (TNF-α), is the prototypic ligand of the TNF superfamily. It is a pleiotropic molecule that plays a central role in inflammation, apoptosis, and immune system development. TNF-α is produced by a wide variety of immune and epithelial cell types. It belongs to the TNF family of ligands and signals through two receptors, TNFR1 and TNFR2. TNF-α is cytotoxic to a wide variety of tumor cells and is an essential factor in mediating the immune response against bacterial infections. TNF-α also plays a role in the induction of septic shock, auto immune diseases, rheumatoid arthritis, inflammation, and diabetes.

Quality

Bioactivity assay: Determined by the dose-dependent cytotoxity of the TNF alpha sensitive cell line L-929 in the presence of
Actinomycin D. Typically <0.9 ng/mL ED50.

Physical form

HumanKine TNF alpha is manufactured using all non-animal reagents and is considered Xeno-Free

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service